Skip to Content
Merck
All Photos(1)

Key Documents

340928

Sigma-Aldrich

1,4-Anhydroerythritol

95%

Synonym(s):

(3R,4S)-Tetrahydro-3,4-furandiol, Erythritol-1,4-anhydride, cis-3,4-Dihydroxytetrahydrofuran, cis-Tetrahydrofuran-3,4-diol, meso-3,4-Dihydroxytetrahydrofuran, rel-(3R,4S)-Tetrahydro-3,4-furandiol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$168.00
25 G
$594.00

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$168.00
25 G
$594.00

About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.477 (lit.)

density

1.27 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

O[C@H]1COC[C@H]1O

InChI

1S/C4H8O3/c5-3-1-7-2-4(3)6/h3-6H,1-2H2/t3-,4+

InChI key

SSYDTHANSGMJTP-ZXZARUISSA-N

General description

1,4-Anhydroerythritol is a cyclic vicinal diol.[1] 1,4-Anhydroerythritol undergoes hydrodeoxygenation over tungsten oxide-palladium catalysts to yield 3-hydroxytetrahydrofuran.[1]

Application

1,4-Anhydroerythritol was employed in a key step in the synthesis of glucopyranoside-based analogs of adenophostin A lacking the adenine component.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasushi Amada et al.
ChemSusChem, 7(8), 2185-2192 (2014-07-01)
Hydrodeoxygenation of cyclic vicinal diols such as 1,4-anhydroerythritol was conducted over catalysts containing both a noble metal and a group 5-7 transition-metal oxide. The combination of Pd and WOx allowed the removal of one of the two OH groups selectively.
Alex C Watanabe et al.
Environmental science & technology, 52(15), 8346-8354 (2018-06-29)
Laboratory and field measurements have demonstrated that isoprene epoxydiol (IEPOX) is the base component of a wide range of chemical species found in isoprene-derived secondary organic aerosol (SOA). To address newly raised questions concerning the chemical identities of IEPOX-derived SOA
Andrew M Riley et al.
Carbohydrate research, 337(12), 1067-1082 (2002-06-14)
Adenophostins A and B are naturally occurring glyconucleotides that interact potently with receptors for D-myo-inositol 1,4,5-trisphosphate, an important second messenger molecule in most cell types. Here we describe the design and synthesis of glucopyranoside-based analogues of adenophostin A lacking the
Aritomo Yamaguchi et al.
Physical chemistry chemical physics : PCCP, 19(4), 2714-2722 (2016-11-30)
The intramolecular dehydration of biomass-derived sugar alcohols d-sorbitol, d-mannitol, galactitol, xylitol, ribitol, l-arabitol, erythritol, l-threitol, and dl-threitol was investigated in high-temperature water at 523-573 K without the addition of any acid catalysts. d-Sorbitol and d-mannitol were dehydrated into isosorbide and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service