Skip to Content
Merck
All Photos(2)

Key Documents

225851

Sigma-Aldrich

Sodium permanganate monohydrate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaMnO4 · H2O
CAS Number:
Molecular Weight:
159.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥95%

form

chunks
crystalline powder

reaction suitability

reagent type: oxidant

SMILES string

O.[Na+].[O-][Mn](=O)(=O)=O

InChI

1S/Mn.Na.H2O.4O/h;;1H2;;;;/q;+1;;;;;-1

InChI key

DJKJCISMVZLEDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetically useful oxidations at solid sodium permanganate surfaces.
Menger FM and Lee C.
Tetrahedron Letters, 22(18), 1655-1656 (1981)
An improved oxidation of an alcohol using aqueous permanganate and phase-transfer catalyst.
Mahmood A, et al.
Organic Process Research & Development, 3(5), 363-364 (1999)
Kathrin M Engel et al.
Chemistry and physics of lipids, 203, 33-45 (2017-01-09)
Oxidized (phospho)lipids are of paramount interest for different reasons: besides their in vivo relevance as markers of inflammatory diseases, they are often needed in the laboratory to study the response of selected cells to oxidized lipids. Mass spectrometry (MS) is
Achieving electrochemical capacitor functionality from nanoscale LiMn2O4 coatings on 3-D carbon nanoarchitectures.
Sassin MB, et al.
Journal of Material Chemistry A, 1(7), 2431-2440 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service