Skip to Content
Merck
All Photos(2)

Key Documents

221732

Sigma-Aldrich

Sodium tetraborate

99%

Synonym(s):

Borax, fused

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7
CAS Number:
Molecular Weight:
201.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Agency

suitable for SM 4500 - NH3

Quality Level

Assay

99%

form

solid

mp

741 °C (lit.)

density

2.367 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1

InChI key

UQGFMSUEHSUPRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium tetraborate (Na2B4O7) can be used as a co-catalyst for the oxidation of alcohols to corresponding carbonyl compounds in greener non-chlorinated solvents in the presence of TEMPO/NaOCl. It is also used as a structure-directing agent as well as a catalyst in the preparation of carbon aerogels using glucose as the carbon precursor.

Legal Information

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zichen Zhao et al.
Foods (Basel, Switzerland), 10(7) (2021-08-08)
UV-B illumination facilitates aggregation of alpha-lactalbumin (α-LA) by intramolecular disulfide bond cleavage followed by intermolecular thiol-disulfide exchange reactions. However, long term exposure to UV-B illumination may induce undesired oxidative modifications of amino acid residues in the protein. The purpose of
Borax-Mediated Formation of Carbon Aerogels from Glucose
Fellinger T-P, et al.
Advances in Functional Materials, 22(15), 3254-3260 (2012)
Towards greener solvents for the bleach oxidation of alcohols catalysed by stable N-oxy radicals
Janssen MHA, et al.
Green Chemistry, 13(4), 905-912 (2011)
Siyu Liu et al.
ACS nano, 14(11), 15936-15943 (2020-11-13)
Flat, membrane-like materials made of graphene oxide (GO) nanoflakes have extraordinary mechanical properties including high stiffness, high strength, and low weight. However, the forming of complex nonplanar structures from flat GO membranes is difficult because of the intrinsic brittleness of
Caio A C G Brunharo et al.
Journal of agricultural and food chemistry, 67(31), 8431-8440 (2019-05-09)
Glufosinate-resistant Lolium perenne L. spp. multiflorum biotypes from Oregon exhibited resistance levels up to 2.8-fold the field rate. One resistant biotype (MG) had an amino acid substitution in glutamine synthetase 2 (GS2), whereas the other (OR) exhibited the wild-type genotype.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service