519073
Sodium permanganate solution
40 wt. % in H2O
Synonym(s):
Sodium manganate, Sodium permanganate (NaMnO4 )
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
liquid
Quality Level
reaction suitability
reagent type: oxidant
concentration
40 wt. % in H2O
technique(s)
electron microscopy: suitable
bp
100 °C
density
1.391 g/mL at 25 °C
SMILES string
[Na+].[O-][Mn](=O)(=O)=O
InChI
1S/Mn.Na.4O/q;+1;;;;-1
InChI key
WPWYHBSOACXYBB-UHFFFAOYSA-N
General description
Sodium permanganate solution (NaMnO4) is a water soluble salt solution that can prepared by using manganese oxide with sodium hypochloride. It can be used as an etchant in electronic circuits. It can also be used as a fixing agent in electron microscopy.
Application
- Describing the permeability of permanganate co-ions in a CEM with varying supporting electrolyte concentrations: Research focused on the transport properties of sodium permanganate in ion-exchange membranes, providing insights into its applications in electrochemical devices. (HJ Cassady, MF Rochow, MA Hickner - Journal of Membrane Science, 2024). Read more.
- The long-term removal of dissolved Mn from drinking water via membrane filtration using sodium permanganate as a pre-oxidant: The study demonstrates the use of sodium permanganate as an effective pre-oxidant in membrane filtration systems for purifying drinking water. (YS Lee, J Eom, IA Khan, JO Kim - Journal of Industrial and Engineering Chemistry, 2024). Read more.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Liq. 2 - Skin Corr. 1B
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cell and tissue fixation, 1972-1982.
The Histochemical journal, 17(4), 389-442 (1985-04-01)
Sodium permanganate fixation for electron microscopy
The Journal of Biophysical and Biochemical Cytology, 9(3), 711-711 (1961)
Selective demonstration of serotonergic neurons and terminals in electron micrographs: loading with 5,7-dihydroxytryptamine and fixation with NaMnO4.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 30(8), 769-773 (1982-08-01)
Journal of contaminant hydrology, 132, 58-74 (2012-03-31)
In situ chemical oxidation (ISCO) treatment with sodium permanganate, an electrically conductive oxidant, provides a strong electrical signal for tracking of injectate transport using time series geophysical surveys including direct current (DC) resistivity and electromagnetic (EM) methods. Effective remediation is
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 52(2), 135-143 (2016-10-22)
Sulfonamide antibiotics are often detected in terrestrial and aquatic environment, but little is known about abiotic degradation of these antibiotics. In the present study, the degradation of the sulfonamide antibiotic sulfadiazine by a synthesized δ-MnO
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service