Skip to Content
Merck
All Photos(3)

Key Documents

244589

Sigma-Aldrich

Manganese(II) chloride

powder and chunks, ≥99% trace metals basis

Synonym(s):

Manganese dichloride, Sacchite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38190105
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99% trace metals basis

form

powder and chunks

impurities

≤10000.0 ppm Trace Metal Analysis

mp

652 °C (lit.)

density

2.98 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Mn]Cl

InChI

1S/2ClH.Mn/h2*1H;/q;;+2/p-2

InChI key

GLFNIEUTAYBVOC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Manganese (II) chloride is a pink crystalline solid that is highly soluble in water. The anhydrous form of Manganese(II) chloride assumes a structure similar to layered cadmium chloride. It is a key starting material for the synthesis of organomanganese compounds.

Application

Manganese(II) chloride can be used:
  • As a precursor to synthesize manganese oxide nanoparticles.
  • To fabricate Mn(ll) coordinated polyaniline electrodes for supercapacitors.
  • To prepare thermally activated high anodic capability MOFs for Li-ion batteries.
  • As a starting material to synthesize polymer composite films withenhanced optical properties.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A thermally activated manganese 1, 4-benzenedicarboxylate metal organic framework with high anodic capability for Li-ion batteries
Huiping Hu, et al.
New Journal of Science, 40, 9746-9752 (2016)
Electrochemical performance of manganese coordinated polyaniline
Yucheng Chen and Yibing Xie
Advanced Electronic Materials, 5, 1900816-1900816 (2019)
Edward Pajarillo et al.
Neurotoxicology, 65, 280-288 (2017-12-01)
Chronic exposure to manganese (Mn) causes neurotoxicity, referred to as manganism, with common clinical features of parkinsonism. 17β-estradiol (E2) and tamoxifen (TX), a selective estrogen receptor modulator (SERM), afford neuroprotection in several neurological disorders, including Parkinson's disease (PD). In the
Microstructural and optical properties of polyvinyl alcohol/manganese chloride composite film
M Banerjee, et al.
Polymer Composites, 40, E765-E775 (2019)
Investigation of the effect of annealing temperature on photophysical properties of manganese dioxide nanostructure prepared via co-precipitation route
M S Iqbal, et al.
IOP Conference Series: Materials Science and Engineering, 683, 012033-012033 (2019)

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service