Skip to Content
Merck
All Photos(1)

Key Documents

1.05927

Supelco

Manganese(II) chloride tetrahydrate

for analysis EMSURE® ACS

Synonym(s):

Manganese(II) chloride tetrahydrate, Manganese dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Mn · 4H2O
CAS Number:
Molecular Weight:
197.91
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-869-6
NACRES:
NA.21
Assay:
99.0-101.0% (complexometric)
grade:
ACS reagent
form:
solid
solubility:
1980 g/L

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.0-101.0% (complexometric)

form

solid

potency

1484 mg/kg LD50, oral (Rat)

impurities

≤0.0005% Substances reducing potassium permanganate (as O)
≤0.005% Insoluble matter
≤0.2% Substances not precipitated by ammonium sulfide (as SO4)

pH

3.5-6 (25 °C, 50 g/L in H2O)

bp

1190 °C/1013 hPa (anhydrous substance)

mp

650 °C (anhydrous substance)

solubility

1980 g/L

density

2.01 g/cm3 at 20 °C

bulk density

1150 kg/m3

anion traces

sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.001%
Mg: ≤0.005%
Na: ≤0.005%
Pb: ≤0.0005%
Zn: ≤0.001%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Mn+2].[Cl-].[Cl-].O.O.O.O

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application



  • High-performance lithium storage in an ultrafine manganese fluoride nanorod anode with enhanced electrochemical activation based on conversion reaction.: Research demonstrates the use of manganese(II) chloride in synthesizing ultrafine manganese fluoride nanorods for high-performance lithium storage, showcasing advancements in battery technology and electrochemical applications (Rui et al., 2016).


  • Synthesis, structural investigation and thermal properties of a novel manganese complex Mn2(DAT)2Cl4(H2O)4 (DAT=1,5-diaminotetrazole).: This paper details the synthesis and characterization of a new manganese complex, contributing to the understanding of manganese chemistry and its potential applications in various industrial processes (Zhang et al., 2010).


  • Poly[tetra-aqua-(μ(6)-9,10-dioxo-9,10-dihydro-anthracene-1,4,5,8-tetra-carboxyl-ato)dimanganese(II)].: The study presents the structural details of a new manganese(II) coordination polymer, providing insights into its potential applications in material science and coordination chemistry (Xu et al., 2012).


  • Metal-organic frameworks based on unprecedented trinuclear and pentanuclear metal-tetrazole clusters as secondary building units.: This research highlights the formation of metal-organic frameworks using manganese(II) chloride, emphasizing its role in developing novel materials with unique structural and functional properties (Jeong et al., 2011).


Analysis Note

Assay (complexometric): 99.0 - 101.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water; 25 °C): 3.5 - 6.0
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.001 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.005 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.001 %
Substances not precipitated by ammonium sulfide (as SO₄): ≤ 0.2 %
Substances reducing potassium permanganate (as O): ≤ 0.0005 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service