Skip to Content
Merck
All Photos(1)

Key Documents

SML1408

Sigma-Aldrich

(R)-PFI-2

≥97% (HPLC)

Synonym(s):

(R)-8-Fluoro-N-(1-oxo-1-(pyrrolidin-1-yl)-3-(3-(trifluoromethyl)phenyl)propan-2-yl)-1,2,3,4-tetrahydroisoquinoline-6-sulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25F4N3O3S · HCl
CAS Number:
Molecular Weight:
535.98
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC1=C2C(CCNC2)=CC(S(N[C@H](CC3=CC=CC(C(F)(F)F)=C3)C(N4CCCC4)=O)(=O)=O)=C1.[H]Cl

InChI

1S/C23H25F4N3O3S.ClH/c24-20-13-18(12-16-6-7-28-14-19(16)20)34(32,33)29-21(22(31)30-8-1-2-9-30)11-15-4-3-5-17(10-15)23(25,26)27;/h3-5,10,12-13,21,28-29H,1-2,6-9,11,14H2;1H/t21-;/m1./s1

InChI key

ZADKZNVAJGEFLC-ZMBIFBSDSA-N

Biochem/physiol Actions

(R)-PFI-2 is a histone-lysine N-methyltransferase (HKMT) inhibitor selective for SETD7 (also known as SET9).
(R)-PFI-2 is a histone-lysine N-methyltransferase (HKMT) inhibitor selective for SETD7 (also known as SET9). (R)-PFI-2 has an IC50 value of 2 nM and 1000-fold selectivity over other methyltransferases and other non-epigenetic targets. For full characterization details, please visit the PFI-2 probe summary on the Structural Genomics Consortium (SGC) website.

(S)-PFI-2, an enantiomer of (R)-PFI-2, is used as a negative control. (S)-PFI-2 is available from Sigma. To learn more about and purchase (S)-PFI-2, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

(R)-PFI-2 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

(R)-PFI-2 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the (R)-PFI-2 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Bichmann et al.
Molecular neurodegeneration, 16(1), 46-46 (2021-07-04)
Human tauopathies including Alzheimer's disease (AD) are characterized by alterations in the post-translational modification (PTM) pattern of Tau, which parallel the formation of insoluble Tau aggregates, neuronal dysfunction and degeneration. While PTMs on aggregated Tau have been studied in detail

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service