Skip to Content
Merck
All Photos(3)

Documents

PZ0017

Sigma-Aldrich

Tofacitinib citrate

≥98% (HPLC), powder, JAK3 inhibitor

Synonym(s):

(3R,4R)-4-Methyl-3-(methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-β-oxo-1-piperidinepropanenitrile citrate salt, 3-[(3R,4R)-4-Methyl-3-[methyl-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]piperidin-1-yl]-3-oxopropanenitrile citrate salt, CP-690550-10, Tasocitinib citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N6O · C6H8O7
CAS Number:
Molecular Weight:
504.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Tofacitinib citrate, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution; warmed)

storage temp.

room temp

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.C[C@@H]1CCN(C[C@@H]1N(C)c2ncnc3[nH]ccc23)C(=O)CC#N

InChI

1S/C16H20N6O.C6H8O7/c1-11-5-8-22(14(23)3-6-17)9-13(11)21(2)16-12-4-7-18-15(12)19-10-20-16;7-3(8)1-6(13,5(11)12)2-4(9)10/h4,7,10-11,13H,3,5,8-9H2,1-2H3,(H,18,19,20);13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/t11-,13+;/m1./s1

InChI key

SYIKUFDOYJFGBQ-YLAFAASESA-N

Gene Information

General description

Tofacitinib is a synthetic molecule corresponding to a molecular weight of 312.4 Da and is permeable by transcellular diffusion.

Application

Tofacitinib citrate has been used:
  • as a ligand for human serum albumin in fluorescence quenching, dynamic light scattering (DLS) measurements, differential scanning calorimetry and molecular docking studies
  • as a medium supplement for full depth articular cartilage (FDC) explants to monitor cytokine-induced proteoglycan loss
  • as a Janus kinase inhibitor in MCF7 breast cancer cells

Biochem/physiol Actions

Tofacitinib is a potent inhibitor of Janus kinase 3 (JAK3) with some JAK-1 inhibitory activity as well. It blocks downstream STAT signaling resulting in potent inhibition of inflammatory cytokines with resultant immunosuppressive and anti-inflammatory activity. Tofacitinib is being investigated for for several autoimmune disorders including, rheumatoid arthritis, psoriasis and dry eye.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

Tofacitinib has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the Tofacitinib probe summary on the Chemical Probes Portal website.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphoproteome profiling reveals critical role of JAK-STAT signaling in maintaining chemoresistance in breast cancer
Nascimento AS, et al.
Testing, 8(70), 114756-114756 (2017)
Ji Sang Lee et al.
Pharmaceutics, 11(7) (2019-07-10)
This study investigated the pharmacokinetics of tofacitinib in rats and the effects of first-pass metabolism on tofacitinib pharmacokinetics. Intravenous administration of 5, 10, 20, and 50 mg/kg tofacitinib showed that the dose-normalized area under the plasma concentration-time curve from time
Tofacitinib and TPCA-1 exert chondroprotective effects on extracellular matrix turnover in bovine articular cartilage ex vivo
Kjelgaard-Petersen CF, et al.
Biochemical Pharmacology (2018)
The mechanism of action of tofacitinib-an oral Janus kinase inhibitor for the treatment of rheumatoid arthritis
Hodge JA, et al.
Clinical and Experimental Rheumatology, 34(2), 318-328 (2016)
Ben X Wang et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 37(4), 147-152 (2017-04-08)
Aicardi-Goutières syndrome (AGS) is an early-onset, genetic disease characterized by recurrent fever, multifocal lesions of the brain, and systemic autoimmunity. We report on 3 AGS patients, 2 siblings with an RNASEH2A gene mutation and 1 patient with a SAMHD1 gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service