Skip to Content
Merck
All Photos(1)

Key Documents

SML1133

Sigma-Aldrich

SGC-CBP30

≥98% (HPLC)

Synonym(s):

2-[2-(3-Chloro-4-methoxyphenyl)ethyl]-5-(3,5-dimethyl-1,2-oxazol-4-yl)-1-[(S)-2-(morpholin-4-yl)propyl]-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H33ClN4O3
CAS Number:
Molecular Weight:
509.04
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(C(C)=NO1)C2=CC=C3C(N=C(CCC4=CC=C(OC)C(Cl)=C4)N3C[C@@H](N5CCOCC5)C)=C2

InChI

1S/C28H33ClN4O3/c1-18(32-11-13-35-14-12-32)17-33-25-8-7-22(28-19(2)31-36-20(28)3)16-24(25)30-27(33)10-6-21-5-9-26(34-4)23(29)15-21/h5,7-9,15-16,18H,6,10-14,17H2,1-4H3/t18-/m0/s1

InChI key

GEPYBHCJBORHCE-SFHVURJKSA-N

Application

SGC-CBP30 has been used in immunofluorescence, to study the regulation of HAT (histone acetyltransferase) activity.[1] It has also been used to abolish synergistic gene expression induced by transcription factors in hepatocytes.[2]

Biochem/physiol Actions

SGC-CBP30 is a selective inhibitor of the bromodomain-containing transcription factors CREBBP (CBP) and EP300 (IC50 = 0.021 and 0.038 μM, respectively). SGC-CBP30 has little activity against other bromodomains at concentrations up to 1 mM. For full characterization details, please visit the SGC-CBP30 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
SGC-CBP30 is a selective inhibitor of the bromodomain-containing transcription factors CREBBP (CBP) and EP300.
SGC-CBP30 might serve as a therapeutic agent in treating cancer.[3] In ankylosing spondylitis and psoriatic arthritis condition, SGC-CBP30 is found to decrease immune cell production of T helper cell 17 and other pro-inflammatory cytokines.[4]

Features and Benefits

SGC-CBP30 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

SGC-CBP30 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the SGC-CBP30 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical progress and pharmacology of small molecule bromodomain inhibitors
Theodoulou NH, et al.
Current Opinion in Chemical Biology, 33, 58-66 (2016)
Epigenetic Targets in Drug Discovery, 144-144 (2018)
Transcription factor dimerization activates the p300 acetyltransferase
Ortega E, et al.
Nature, 1-1 (2018)
Synergistic gene expression during the acute phase response is characterized by transcription factor assisted loading.
Goldstein I, et al.
Nature Communications, 8(1), 1849-1849 (2017)
Kee-Beom Kim et al.
Science advances, 8(7), eabl4618-eabl4618 (2022-02-17)
EP300, a transcription coactivator important in proliferation and differentiation, is frequently mutated in diverse cancer types, including small cell lung cancer (SCLC). While these mutations are thought to result in loss of EP300 function, the impact on tumorigenesis remains largely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service