Skip to Content
Merck
All Photos(1)

Key Documents

SML1242

Sigma-Aldrich

SGC707

≥98% (HPLC)

Synonym(s):

1-Isoquinolin-6-yl-3-(2-oxo-2-pyrrolidin-1-yl-ethyl)-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O2
CAS Number:
Molecular Weight:
298.34
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCC(N1CCCC1)=O)NC2=CC3=CC=NC=C3C=C2

InChI

1S/C16H18N4O2/c21-15(20-7-1-2-8-20)11-18-16(22)19-14-4-3-13-10-17-6-5-12(13)9-14/h3-6,9-10H,1-2,7-8,11H2,(H2,18,19,22)

InChI key

DMIDPTCQPIJYFE-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

SGC707 is a potent allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3).
SGC707 is a potent allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3). SGC707 has an IC50 value of 50 nM and >100-fold selectivity over other methyltransferases and non-epigenetic targets. SGC707 binds to PRMT3 with KD of 50 nM (ITC), and inhibits histone methylation in cells with an IC50 value below 1 μM. For full characterization details, please visit the SGC707 probe summary on the Structural Genomics Consortium (SGC) website.

XY1 is the negative control for the active probe, SGC707. To request a sample of the negative control from the SGC, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

SGC707 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

SGC707 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the SGC707 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service