Skip to Content
Merck
All Photos(1)

Key Documents

SML1209

Sigma-Aldrich

OICR-9429

≥98% (HPLC)

Synonym(s):

6-Oxo-4-trifluoromethyl-1,6-dihydro-pyridine-3-carboxylic acid [4-(4-methyl-piperazin-1-yl)-3′-morpholin-4-ylmethyl-biphenyl-3-yl]-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H32F3N5O3
CAS Number:
Molecular Weight:
555.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CN(C(=O)NC1=O)CC#C

InChI

1S/C7H5FN2O2/c1-2-3-10-4-5(8)6(11)9-7(10)12/h1,4H,3H2,(H,9,11,12)

InChI key

GMCOGJQJFDQZCP-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

OICR-9429 is a cell penetrant, potent and selective antagonist of the interaction of WDR5 (WD repeat domain 5) with peptide regions of MLL and Histone 3 that potently binds to WDR5.
OICR-9429 is a cell penetrant, potent and selective antagonist of the interaction of WDR5 (WD repeat domain 5) with peptide regions of MLL and Histone 3 that potently binds to WDR5. OICR-9429 inhibits the interaction of WDR5 with MLL1 and RbBP5 in cells. For full characterization details, please see OICR-9429 on the Structural Genomics Consortium (SGC) website.

OICR-0547 is the negative control for the active probe, OICR-9429. To request a sample of the negative control from the SGC, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

OICR-9429 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

OICR-9429 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the OICR-9429 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service