Skip to Content
Merck
All Photos(2)

Key Documents

04367

Sigma-Aldrich

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

≥98.0% (T)

Synonym(s):

[EMIM][TRIFLATE], EMIM Otf

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11F3N2O3S
CAS Number:
Molecular Weight:
260.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

liquid

impurities

≤0.1% water

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

>350 °C (lit.)

density

1.387 g/mL at 25 °C (lit.)

SMILES string

CCn1cc[n+](C)c1.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H11N2.CHF3O3S/c1-3-8-5-4-7(2)6-8;2-1(3,4)8(5,6)7/h4-6H,3H2,1-2H3;(H,5,6,7)/q+1;/p-1

InChI key

ZPTRYWVRCNOTAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate may be used as a solvent to produce Ionic Polymer-Polymer Composites (IP2C) and also in lipase-catalyzed enantioselective amine acylation with 4-pentenoic acid.

Other Notes

Ionic liquid stable to air and water useful for electrochemical purposes (molten salt technique) exhibiting an electrochemical stability window of ≥4 V and being stable to up to 350 C

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

425.3 °F - closed cup

Flash Point(C)

218.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.I. Cooper et al.
Proc. Electrochem. Soc., 16, 386-386 (1992)
Heat capacities and excess enthalpies of 1-ethyl-3-methylimidazolium-based ionic liquids and water
Ficke, Lindsay E., He?ctor Rodri?guez, and Joan F. Brennecke
Journal of Chemical and Engineering Data, 53.9, 2112-2119 (2008)
Capillary electrophoretic application of 1-alkyl-3-methylimidazolium-based ionic liquids
Yanes, Enrique G., et al.
Analytical Chemistry, 73.16, 3838-3844 (2001)
A study on IP2C actuators using ethylene glycol or EmI-Tf as solvent.
Di Pasquale G, et al.
Smart Materials and Structures, 20(4), 045014-045014 (2011)
Investigation of ionic liquids as reaction media for enzymatic enantioselective acylation of amines.
Irimescu R and Kato K.
Journal of Molecular Catalysis. B, Enzymatic, 30(5), 189-194 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service