Skip to Content
Merck
All Photos(1)

Key Documents

00768

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tetrafluoroborate

for electrochemistry, ≥99.0% (HPLC)

Synonym(s):

EMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BF4N2
CAS Number:
Molecular Weight:
197.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for electrochemistry

Quality Level

Assay

≥99.0% (HPLC)

form

liquid

impurities

≤200 ppm water

refractive index

n20/D 1.413 (lit.)

bp

>350 °C (lit.)

mp

15 °C (lit.)

density

1.294 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

F[B-](F)(F)F.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.BF4/c1-3-8-5-4-7(2)6-8;2-1(3,4)5/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

CUNYTRQQXKCRTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Ethyl-3-methylimidazolium tetrafluoroborate is a room temperature ionic liquid widely used as an electrolyte in electrochemical studies. Its diffusion coefficient, 13C spin-lattice relaxation times, and nuclear Overhauser enhancements have been analyzed using NMR techniques to study its hydrogen bonding. Some of its fundamental physical and chemical properties such as density, viscosity, thermal stability, surface tension, refractive index and pH have been studied over a wide range of temperature.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical and electrochemical properties of 1-alkyl-3-methylimidazolium tetrafluoroborate for electrolyte
Nishida T, et al.
Journal of Fluorine Chemistry, 120(2), 135-141 (2003)
Electrochemical study of copper in a basic 1-ethyl-3-methylimidazolium tetrafluoroborate room temperature molten salt
Chen PY and Sun IW
Electrochimica Acta, 45(3), 441-450 (1999)
NMR evidence of hydrogen bonding in 1-ethyl-3-methylimidazolium-tetrafluoroborate room temperature ionic liquid
Huang JF, et al.
Inorgorganica Chimica Acta, 320(1-2), 7-11 (2001)
The Room Temperature Ionic Liquid 1-Ethyl-3-methylimidazolium Tetrafluoroborate: Electrochemical Couples and Physical Properties.
Fuller J, et al.
Journal of the Electrochemical Society, 144(11), 3881-3886 (1997)
Sungwon Lee et al.
Nature communications, 5, 5898-5898 (2014-12-20)
To measure electrophysiological signals from the human body, it is essential to establish stable, gentle and nonallergic contacts between the targeted biological tissue and the electrical probes. However, it is difficult to form a stable interface between the two for

Articles

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service