Skip to Content
Merck
All Photos(1)

Key Documents

77896

Sigma-Aldrich

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

≥98.0% (HPLC)

Synonym(s):

BMIIm, BMIM TFSI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15F6N3O4S2
CAS Number:
Molecular Weight:
419.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.428

functional group

fluoro

SMILES string

CCCCn1cc[n+](C)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C8H15N2.C2F6NO4S2/c1-3-4-5-10-7-6-9(2)8-10;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h6-8H,3-5H2,1-2H3;/q+1;-1

InChI key

INDFXCHYORWHLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is a room temperature ionic liquid (RTIL). The solubility of alkane gases are less than the alkene gas in this IL. Its ability to solubilize CO2 is more when compared to 1-butyl-3-methylimidazolium dicyanamide.

Application

  • Application of 1-butyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide ionic liquid for different types of separations problem: Focuses on its application in separation processes, highlighting its environmental benefits and effectiveness (S Singh et al., 2016, Elsevier).

Physical form

ionic liquid

Legal Information

Product protected by U.S. Patent 5,827,602 assigned to Covalent Associates, Inc. Woburn, MA, USA

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The gaseous enthalpy of formation of the ionic liquid 1-butyl-3-methylimidazolium dicyanamide from combustion calorimetry, vapor pressure measurements, and ab initio calculations
Emel'yanenko, Vladimir N., Sergey P. Verevkin, and Andreas Heintz
Journal of the American Chemical Society, 129.13, 3930-3937 (2007)
Speciation of Uranyl Nitrato Complexes in Acetonitrile and in the Ionic Liquid 1?Butyl?3?methylimidazolium Bis (trifluoromethylsulfonyl) imide
Servaes, Kelly, et al.
European Journal of Inorganic Chemistry, 2007.32, 5120-5126 (2007)
A comparative study on the reactivity of electrogenerated bromine with cyclohexene in acetonitrile and the room temperature ionic liquid, 1-butyl-3-methylimidazolium bis [(trifluoromethyl) sulfonyl] imide
Allen, Gary D., et al.
The Journal of Physical Chemistry B, 108.41, 16322-16327 (2004)
Physicochemical properties, structure, and conformations of 1-butyl-3-methylimidazolium bis (trifluoromethanesulfonyl) imide [C4mim]NTf2 ionic liquid.
Blokhin AV, et al.
The Journal of Physical Chemistry B, 112(14), 4357-4364 (2008)
Solubilities of gases in the ionic liquid 1-n-butyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide.
Lee BC and Outcalt SL.
Journal of Chemical and Engineering Data, 51(3), 892-897 (2006)

Articles

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service