Skip to Content
Merck
All Photos(2)

Key Documents

711721

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tetrafluoroborate

≥98% (HPLC)

Synonym(s):

EMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BF4N2
CAS Number:
Molecular Weight:
197.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98% (HPLC)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.413 (lit.)

bp

>350 °C (lit.)

mp

15 °C (lit.)

density

1.294 g/mL at 25 °C (lit.)

SMILES string

F[B-](F)(F)F.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.BF4/c1-3-8-5-4-7(2)6-8;2-1(3,4)5/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

CUNYTRQQXKCRTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium tetrafluoroborate is an air and water stable, room temperature ionic liquid (RTIL). It can be prepared by reacting 1-ethyl-3-methylimidazolium bromide with tetrafluoroboric acid.

Application


  • 1-Ethyl-3-methylimidazolium tetrafluoroborate-doped high ionic conductivity gel electrolytes with reduced anodic reaction potentials for electrochromic devices: This study reports the preparation of a 1-Ethyl-3-methylimidazolium tetrafluoroborate ([Emim] BF4)-doped Poly(methyl methacrylate)(PMMA)-based gel electrolyte for electrochromic applications, showing improved ionic conductivity and reduced anodic reaction potentials (Tang et al., 2017).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flexible Asymmetric Supercapacitor Based on Structure?Optimized Mn3O4/Reduced Graphene Oxide Nanohybrid Paper with High Energy and Power Density
Hu, Yating, et al.
Advances in Functional Materials, 25.47, 7291-7299 (2015)
Solubility of carbon dioxide in 1-ethyl-3-methylimidazolium tetrafluoroborate
Soriano, Allan N., Bonifacio T. Doma Jr, and Meng-Hui Li
Journal of Chemical and Engineering Data, 53.11, 2550-2555 (2008)
Crystal structures of frozen room temperature ionic liquids, 1-ethyl-3-methylimidazolium tetrafluoroborate (EMImBF 4), hexafluoroniobate (EMImNbF 6) and hexafluorotantalate (EMImTaF 6), determined by low-temperature X-ray diffraction
Matsumoto, Kazuhiko, et al.
Solid State Sciences, 8.10, 1250-1257 (2006)
Efficient, halide free synthesis of new, low cost ionic liquids: 1, 3-dialkylimidazolium salts containing methyl-and ethyl-sulfate anions
Holbrey, John D., et al.
Green Chemistry, 4.5, 407-413 (2002)
NMR evidence of hydrogen bonding in 1-ethyl-3-methylimidazolium-tetrafluoroborate room temperature ionic liquid
Huang, Jing-Fang, et al.
Inorgorganica Chimica Acta, 320.1, 7-11 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service