Skip to Content
Merck
All Photos(1)

Key Documents

43437

Sigma-Aldrich

1-Ethyl-3-methylimidazolium thiocyanate

≥95% (H-NMR, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3S
CAS Number:
Molecular Weight:
169.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95% (H-NMR, HPLC)

impurities

≤1% water

mp

-6 °C

SMILES string

[S-]C#N.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.CHNS/c1-3-8-5-4-7(2)6-8;2-1-3/h4-6H,3H2,1-2H3;3H/q+1;/p-1

InChI key

VASPYXGQVWPGAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service