Skip to Content
Merck
All Photos(1)

Key Documents

711756

Sigma-Aldrich

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

≥98% (H-NMR)

Synonym(s):

EMIM Otf

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11F3N2O3S
CAS Number:
Molecular Weight:
260.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (H-NMR)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.435 (lit.)

bp

>350 °C (lit.)

density

1.387 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

SMILES string

CCn1cc[n+](C)c1.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H11N2.CHF3O3S/c1-3-8-5-4-7(2)6-8;2-1(3,4)8(5,6)7/h4-6H,3H2,1-2H3;(H,5,6,7)/q+1;/p-1

InChI key

ZPTRYWVRCNOTAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate may be used as a solvent to produce ionic polymer-polymer composites (IP2C) and also in lipase-catalyzed enantioselective amine acylation with 4-pentenoic acid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

425.3 °F - closed cup

Flash Point(C)

218.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion-pair structure of vaporized ionic liquid studied by matrix-isolation FTIR spectroscopy with DFT calculations: A case of 1-ethyl-3-methylimidazolium trifluoromethanesulfonate
Akai, Nobuyuki, Akio Kawai, and Kazuhiko Shibuya
The Journal of Physical Chemistry A, 114.48, 12662-12666 (2010)
Dielectric relaxation and underlying dynamics of acetonitrile and 1-ethyl-3-methylimidazolium triflate mixtures using THz transmission spectroscopy
Asaki, M. L. T., et al.
J. Chem. Phys. , 116.23, 10377-10385 (2002)
Heat capacities and excess enthalpies of 1-ethyl-3-methylimidazolium-based ionic liquids and water
Ficke, Lindsay E., He?ctor Rodri?guez, and Joan F. Brennecke
Journal of Chemical and Engineering Data, 53.9, 2112-2119 (2008)
A study on IP2C actuators using ethylene glycol or EmI-Tf as solvent.
Di Pasquale G, et al.
Smart Materials and Structures, 20(4), 045014-045014 (2011)
Capillary electrophoretic application of 1-alkyl-3-methylimidazolium-based ionic liquids
Yanes, Enrique G., et al.
Analytical Chemistry, 73.16, 3838-3844 (2001)

Articles

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service