Skip to Content
Merck
All Photos(2)

Key Documents

850355P

Avanti

16:0 PC (DPPC)

Avanti Research - A Croda Brand

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphocholine; DPPC; PC(16:0/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dipalmitoyl-sn-glycero-3-phosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850355P-1g)
pkg of 1 × 200 mg (850355P-200mg)
pkg of 1 × 25 mg (850355P-25mg)
pkg of 1 × 500 mg (850355P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C40H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-39(42)46-36-38(37-48-50(44,45)47-35-34-41(3,4)5)49-40(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3

InChI key

KILNVBDSWZSGLL-UHFFFAOYSA-N

General description

1,2-dihexadecanoyl-sn-glycero-3-phosphocholine (DPPC) is synthesized from unsaturated phosphatidylcholine (PC) by direct de novo synthesis via α1,2-dipalmitoyl diacylglycerol intermediate. The enzyme cholinephosphotransferase catalyzes the conversion of 1,2-dipalmitoyl diacylglycerol to DPPC. Synthesis of DPPC also occurs via remodeling pathway involving deacylation of unsaturated PC.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

16:0 PC (DPPC) has been used in the generation of thymoquinone (TQ) liposomes, gold nanorods and calcium chloride encapsulated liposomes and bilayers for biophysical studies.

Biochem/physiol Actions

1,2-dihexadecanoyl-sn-glycero-3-phosphocholine (DPPC) is a surfactant and reduces surface tension. It is used in combination with 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) for lipid bilayer generation. DPPC alone and in combination with other phospholipid is effective in drug encapsulation and delivery.

Packaging

20 mL Clear Glass Screw Cap Vial (850355P-1g)
20 mL Clear Glass Screw Cap Vial (850355P-500mg)
5 mL Amber Glass Screw Cap Vial (850355P-200mg)
5 mL Amber Glass Screw Cap Vial (850355P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and characterization of a lysophosphatidylcholine acyltransferase in alveolar type II cells
Chen X, et al.
Proceedings of the National Academy of Sciences of the USA, 103(31), 11724-11729 (2006)
Phenotypic basis for matrix stiffness-dependent chemoresistance of breast cancer cells to doxorubicin
Joyce MH, et al.
Frontiers in Oncology, 8, 203-208 (2018)
M Mamusa et al.
Langmuir : the ACS journal of surfaces and colloids, 34(30), 8952-8961 (2018-07-07)
The emergence of antibiotic-resistant bacterial strains has fostered fundamental research to develop alternative antimicrobial strategies. Among the several systems proposed so far, the association complexes (nanoplexes) formed by transcription factor decoys (TFDs), i.e., short oligonucleotides targeting a crucial bacterial transcription
Norbert Kučerka et al.
Langmuir : the ACS journal of surfaces and colloids, 33(12), 3134-3141 (2017-03-10)
Interactions of calcium (Ca2+) and zinc (Zn2+) cations with biomimetic membranes made of dipalmitoylphosphatidylcholine (DPPC) were studied by small angle neutron diffraction (SAND). Experiments show that the structure of these lipid bilayers is differentially affected by the two divalent cations.
Bao Fu Qiao et al.
The journal of physical chemistry. B, 117(17), 5073-5080 (2013-04-10)
Crystalline vesicles are promising nanomaterials due to their mechanical stability in various environments. To control their fabrication, it is essential to understand the effects of different experimental conditions on crystallization. Here we perform atomistic molecular dynamics simulations of anionic lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service