Skip to Content
Merck
All Photos(2)

Key Documents

840035C

Avanti

18:1 PS (DOPS)

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); DOPS; PS(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H77NO10PNa
CAS Number:
Molecular Weight:
810.03
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1,2-dioleoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840035C-10mg)
pkg of 1 × 2.5 mL (840035C-25mg)
pkg of 2 × 4 mL (840035C-200mg)
pkg of 5 × 4 mL (840035C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840035C-10mg)
10 mg/mL (840035C-25mg)
25 mg/mL (840035C-200mg)
25 mg/mL (840035C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C42H78NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(44)50-35-38(36-51-54(48,49)52-37-39(43)42(46)47)53-41(45)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,38-39H,3-16,21-37,43H2,1-2H3,(H,46,47)(H,48,49);/q;+1/p-1/b19-17-,20-18-;/t38-,39+;/m1./s1

InChI key

KPHZNDUWYZIXFY-YORIBCANSA-M

Related Categories

General description

DOPS is an excellent substitute for the naturally occurring Brain PS. It has similar physical properties and is more stable to oxidation. DOPS has been used, along with DOPC and DOPE, in lipid mixtures to mimic platelet membranes for coagulation studies.
Phosphoserine/phosphatidylserine (PS) is an endogenous phospholipid,[1] that is found in all polyanionic macromolecules.[2] 1,2-dioleoyl-sn-glycero-3-phospho-L-serine (DOPS) carry a carboxylic acid group and a protonated amine group in their head group.[3]

Application

18:1 PS (DOPS) has been used as a component of supported membrane tubes (SMrT) templates.[4] It has also been used in the preparation of lipid-mixing vesicles, content-mixing vesicles and lipid mixture.[5]

Biochem/physiol Actions

Phosphoserine (PS) can be used as a template for the recognition of cancer antigen 125 (CA 125).[6] It is very essential for healthy nerve cell membranes and myelin.[1] Exogenous PS can be actively absorbed in humans. It plays a key role in the formation of short-term memory, the consolidation of long-term memory, the ability to create new memories and several other cognitive activities. It actively participates in the locomotor functions, like rapid reactions and reflexes.[1]

Packaging

30 mL Amber Narrow Mouth Bottle with Screw Cap (840035C-500mg)
5 mL Clear Glass Sealed Ampule (840035C-10mg)
5 mL Clear Glass Sealed Ampule (840035C-200mg)
5 mL Clear Glass Sealed Ampule (840035C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A single vesicle-vesicle fusion assay for in vitro studies of SNAREs and accessory proteins
Diao J, et al.
Nature Protocols, 7(5), 921-921 (2012)
Use of the supported membrane tube assay system for real time analysis of membrane fission reactions
Dar S, et al.
Nature Protocols, 12(2), 390-390 (2017)
Choosing the right fluorophore for single-molecule fluorescence studies in a lipid environment
Zhang Z, et al.
Biochimica et Biophysica Acta - Biomembranes, 1859(7), 1242- 1253 (2017)
Toshihiko Oka et al.
Langmuir : the ACS journal of surfaces and colloids, 33(43), 12487-12496 (2017-10-03)
Electrostatic interactions (EIs) play important roles in the structure and stability of inverse bicontinuous cubic (QII) phases of lipid membranes. We examined the effect of pH on the phase of dioleoylphosphatidylserine (DOPS)/monoolein (MO) membranes at low ionic strengths using small-angle
Adsorption of O-phospho-L-serine and L-serine onto poorly crystalline apatite
Benaziz L, et al.
Journal of Colloid and Interface Science, 238(1), 48-53 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service