Skip to Content
Merck
All Photos(1)

Key Documents

880126P

Avanti

DSPE-PEG(2000) Maleimide

Avanti Research - A Croda Brand 880126P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[maleimide(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 10 mg (880126P-10mg)
pkg of 1 × 100 mg (880126P-100mg)
pkg of 1 × 25 mg (880126P-25mg)
pkg of 1 × 50 mg (880126P-50mg)
pkg of 1 × 500 mg (880126P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880126P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\NC(CCN(C(C=C1)=O)C1=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C

InChI

1S/C53H94N3O57P.24C2H4.CH4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-51(60)66-45-47(71-52(61)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)46-70-114(63,64)69-42-39-55-53(62)65-43-44-68-73-75-77-79-81-83-85-87-89-91-93-95-97-99-101-103-105-107-109-11

Application

DSPE-PEG(2000) maleimide is suitable for conjugation with photon-sensitive cell penetrating peptides (psCPP) to prepare nanostructured lipid carrier.[1] It is also suitable for non-covalently functionalizing oxidized multi-walled carbon nanotubes (O-MWCNs).[2]

Biochem/physiol Actions

DSPE-PEG(2000) maleimide is used to prepare conjugated polymer nanoparticles.[3] It is suitable for its use in the synthesis of functional conjugates.[4]

Packaging

20 mL Clear Glass Screw Cap Vial (880126P-500mg)
5 mL Amber Glass Screw Cap Vial (880126P-100mg)
5 mL Amber Glass Screw Cap Vial (880126P-10mg)
5 mL Amber Glass Screw Cap Vial (880126P-25mg)
5 mL Amber Glass Screw Cap Vial (880126P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The targeted delivery of anticancer drugs to brain glioma by PEGylated oxidized multi-walled carbon nanotubes modified with angiopep-2
Ren J, et al.
Biomaterials, 33(11), 3324-3333 (2012)
Conjugated Polymers for Biological and Biomedical Applications, 29-29 (2018)
Yen Ling Yeow et al.
EMBO molecular medicine, 11(12), e10923-e10923 (2019-11-12)
High extracellular matrix (ECM) content in solid cancers impairs tumour perfusion and thus access of imaging and therapeutic agents. We have devised a new approach to degrade tumour ECM, which improves uptake of circulating compounds. We target the immune-modulating cytokine
Araz Norouz Dizaji et al.
Journal of biomaterials science. Polymer edition, 31(3), 293-309 (2019-11-26)
This study aims to develop fluorescence labelled polymeric nanoparticle (NP) carrying vancomycin as the targeting agent for in vivo imaging of Methicillin-resistant Staphylococcus aureus bacterial infections in animal models. Maleimide functionalized 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[maleimide (polyethylene glycol)-2000] as the main was carrier matrix to
Thomas Linemann et al.
Pharmaceutics, 5(2), 246-260 (2013-12-05)
The aim of the present study was to evaluate the transfection potential of chitosan-coated, green-fluorescent magnetic nanoparticles (MNPs) (chi-MNPs) after encapsulation inside polyethylglycol (PEG)ylated liposomes that produced lipid-encapsulated chitosan-coated MNPs (lip-MNPs), and also to evaluate how these particles would distribute

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service