Skip to Content
Merck
All Photos(2)

Key Documents

850757P

Avanti

16:0-18:1 PE

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine; POPE; PE(16:0/18:1(9Z)); 110637

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76NO8P
CAS Number:
Molecular Weight:
718.00
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850757P-1g)
pkg of 2 × 100 mg (850757P-200mg)
pkg of 5 × 100 mg (850757P-500mg)
pkg of 1 × 25 mg (850757P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H76NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,37H,3-16,19-36,40H2,1-2H3,(H,43,44)/b18-17-

InChI key

FHQVHHIBKUMWTI-ZCXUNETKSA-N

Application

16:0-18:1 PE has been used for the preparation of large unilamellar vesicles (LUV).[1][2] It has also been used for the preparation of liposomes for dual polarisation interferometry (DPI).[3]

Packaging

5 mL Clear Glass Sealed Ampule (850757P-200mg)
5 mL Clear Glass Sealed Ampule (850757P-25mg)
5 mL Clear Glass Sealed Ampule (850757P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (850757P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of liprotides to stabilize and transport hydrophobic molecules
Pedersen JN, et al.
Biochemistry, 54(31), 4815-4823 (2015)
Using protein-fatty acid complexes to improve vitamin D stability
Pedersen JN, et al.
Journal of Dairy Science, 99(10), 7755-7767 (2016)
Examination of the Interaction between a Membrane Active Peptide and Artificial Bilayers by Dual Polarisation Interferometry
Payne JAE, et al.
Bio-protocol, 7(1), e2087-e2087 (2017)
Mariya Chavarha et al.
Langmuir : the ACS journal of surfaces and colloids, 28(48), 16596-16604 (2012-11-13)
Prior studies have shown that the biological mixture of the two hydrophobic surfactant proteins, SP-B and SP-C, produces faster adsorption of the surfactant lipids to an air/water interface, and that they induce 1-palmitoyl-2-oleoyl phosphatidylethanolamine (POPE) to form inverse bicontinuous cubic
Charles G Cranfield et al.
Langmuir : the ACS journal of surfaces and colloids, 33(26), 6630-6637 (2017-06-14)
Cyclotides are cyclic disulfide-rich peptides that are chemically and thermally stable and possess pharmaceutical and insecticidal properties. The activities reported for cyclotides correlate with their ability to target phosphatidylethanolamine (PE)-phospholipids and disrupt cell membranes. However, the mechanism by which this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service