Skip to Content
Merck
All Photos(1)

Key Documents

700100P

Avanti

Cholesterol (plant)

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-en-3β-ol; 110797

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

description

cholesterol (plant derived)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (700100P-1g)
pkg of 1 × 100 mg (700100P-100mg)
pkg of 1 × 200 mg (700100P-200mg)
pkg of 1 × 500 mg (700100P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

General description

Cholesterol is a minor sterol present in plants. It is majorly associated with the plant membranes and is a constituent of leaf surface lipids.[1]

Application

Cholesterol has been used:
  • in the preparation of small unilamellar vesicles[2]
  • in the generation of thymoquinone (TQ) loaded liposomes for administering into murine model with Candida albicans infection[3]
  • in the preparation of small unilamellar vesicles for structured illumination microscopy studies[4]

Packaging

20 mL Clear Glass Screw Cap Vial (700100P-1g)
20 mL Clear Glass Screw Cap Vial (700100P-500mg)
5 mL Amber Glass Screw Cap Vial (700100P-100mg)
5 mL Amber Glass Screw Cap Vial (700100P-200mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preferential interactions of primary amine-terminated quantum dots with membrane domain boundaries and lipid rafts revealed with nanometer resolution
Mensch AC, et al.
Environmental science. Nano (2020)
Imad Younus Hasan et al.
Analytical chemistry, 88(10), 5037-5041 (2016-05-04)
Understanding the distribution of cholesterol in phospholipid membranes is of key importance in membrane biophysics, primarily since cholesterol enriched regions, rafts, are known to play a special role in protein function. In this work, quartz crystal microbalance with dissipation (QCM)-based
Cholesterol and plants
Behrman EJ and Gopalan V
Journal of Chemical Education, 82(12), 1791-1791 (2005)
Maheshinie Rajapaksha et al.
Biochemistry, 52(41), 7242-7253 (2013-09-24)
Although the mechanism by which the steroidogenic acute regulatory protein (StAR) promotes steroidogenesis has been studied extensively, it remains incompletely characterized. Because structural analysis has revealed a hydrophobic sterol-binding pocket (SBP) within StAR, this study sought to examine the regulatory
Roy Ziblat et al.
Biophysical journal, 103(2), 255-264 (2012-08-03)
Grazing incidence x-ray diffraction measurements were performed on single hydrated bilayers and monolayers of Ceramide/Cholesterol/1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocyholine at varying concentrations. There are substantial differences in the phase and structure behavior of the crystalline domains formed within the bilayers relative to the corresponding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service