Skip to Content
Merck
All Photos(2)

Key Documents

850345P

Avanti

14:0 PC (DMPC)

1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1,2-ditetradecanoyl-sn-glycero-3-phosphocholine; DMPC; PC(14:0/14:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H72NO8P
CAS Number:
Molecular Weight:
677.93
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850345P-1g)
pkg of 1 × 200 mg (850345P-200mg)
pkg of 1 × 25 mg (850345P-25mg)
pkg of 1 × 500 mg (850345P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850345P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O)=O

InChI

1S/C36H72NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-35(38)42-32-34(33-44-46(40,41)43-31-30-37(3,4)5)45-36(39)29-27-25-23-21-19-17-15-13-11-9-7-2/h34H,6-33H2,1-5H3/t34-/m0/s1

InChI key

CITHEXJVPOWHKC-UMSFTDKQSA-N

General description

Dimyristoylphosphatidylcholine (DMPC) belongs to the phosphatidylcholine (PCs) family. It is composed of saturated and symmetric phospholipids with 14 carbon atoms. DMPC is zwitterionic glycerophospholipid and is associated with mammalian membranes.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

14:0 PC (DMPC) has been used:
  • in the preparation of liposomes for dissipation monitoring and perforin binding and assembly by Quartz crystal microscopy studies.
  • in anthraquinone liposomes generation for spectral studies
  • in bicelles preparation along with phosphatidylinositol 4,5-bisphosphate for in vitro reconstitution of β2-adrenergic receptor

Biochem/physiol Actions

Dimyristoylphosphatidylcholine (DMPC) in combination with cholesterol is used as biomembrane model. Oral supplementation of DMPC in apolipoprotein (apo) E−null mice, improves high density lipoprotein levels and other lipid profiles, preventing the pathogenesis of atherosclerosis.

Packaging

20 mL Clear Glass Screw Cap Vial (850345P-1g)
20 mL Clear Glass Screw Cap Vial (850345P-500mg)
5 mL Amber Glass Screw Cap Vial (850345P-200mg)
5 mL Amber Glass Screw Cap Vial (850345P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oral synthetic phospholipid (DMPC) raises high-density lipoprotein cholesterol levels, improves high-density lipoprotein function, and markedly reduces atherosclerosis in apolipoprotein E-null mice
Navab M, et al.
Circulation, 108(14), 1735-1739 (2003)
Analysis of perforin assembly by quartz crystal microbalance reveals a role for cholesterol and calcium-independent membrane binding
Stewart SE, et al.
Test, 290(52), 31101-31112 (2015)
A coarse-grained molecular dynamics study of DLPC, DMPC, DPPC, and DSPC mixtures in aqueous solution
Abedi K, et al.
Journal of Chemistry, 2013 (2013)
Assembly of streptolysin O pores assessed by quartz crystal microbalance and atomic force microscopy provides evidence for the formation of anchored but incomplete oligomers
Stewart SE, et al.
Biochimica et Biophysica Acta - Biomembranes, 1848(1), 115-126 (2015)
Structural and functional analysis of a beta2-adrenergic receptor complex with GRK5
Komolov KE, et al.
Cell, 169(3), 407-421 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service