Skip to Content
Merck
All Photos(1)

Key Documents

08982

Supelco

Sodium metabisulfite

analytical standard

Synonym(s):

E223, Sodium disulfite, Sodium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O5
CAS Number:
Molecular Weight:
190.11
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E223
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (iodometric, RT)

shelf life

limited shelf life, expiry date on the label

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
thiosulfate (S2O32-): ≤500 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

HRZFUMHJMZEROT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium metabisulfite is primarily used as an antioxidant and antimicrobial agent in a wide variety of foods and drugs. It is also used as the main additive in the prevention of melanosis in shrimp.

Application

Sodium metabisulfite may be used as an antioxidant which when added to anesthetic formulations serves to minimize epinephrine oxidation in the quantification of epinephrine in dental anesthetic solutions by capillary electrophoresis. Sodium metabisulfite may also be used as a standard in the determination of catecholamines in human plasma by high-performance liquid chromatography with electrochemical detection.

Analysis Note

metal trace analysis: corresponds to requirements

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of catecholamines in human plasma by high-performance liquid chromatography with electrochemical detection.
Raggi MA, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 730(2), 201-211 (1999)
The molecular mechanisms of sodium metabisulfite on the expression of KATP and L-Ca2+ channels in rat hearts.
Zhang Q, et al.
Regulatory Toxicology and Pharmacology, 72(3), 440-446 (2015)
Quantitative assay for epinephrine in dental anesthetic solutions by capillary electrophoresis.
Kranack A and Chen DY
Analyst, 123(7), 1461-1463 (1998)
Standardization of sodium metabisulfite solution concentrations and immersion time for farmed shrimp Litopenaeus vannamei.
Andrade LTD, et al.
Ciencia Rural, 45(3), 499-504 (2015)
Rosie F Davies et al.
Clinics in dermatology, 29(3), 311-315 (2011-04-19)
Human skin is exposed to a large variety of cosmetic allergens. Most allergic contact dermatitis occurs after exposure to fragrance, preservatives, and hair dyes. Such reactions can often be occult. As a result, a high index of suspicion is needed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service