Skip to Content
Merck
All Photos(1)

Documents

00180585

Hyperoside

primary reference standard

Synonym(s):

Quercetin 3-D-galactoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-D-galactoside, Hyperin, Hyperoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Beilstein:
5784795
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15+,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-DTGCRPNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hyperoside is a flavonol glycoside compound used in Traditional Chinese medicine for its various beneficial effects, such as anti-inflammatory, anti-oxidant, anti-viral, and anti-cancer.
Produced and qualified by HWI pharma services GmbH.
Exact content measured by quantitative NMR can be found on the certificate.

Application

The primary reference standard can be used for identification, assay, and purity testing, method development, and validation.

It can also be used as follows:

  • Development of an ultra performance liquid chromatography-mass spectrometric method to determine hyperoside in rat plasma samples following sample treatment by protein precipitation and liquid-liquid extraction
  • Multi-residue analysis of hyperoside, isoquercitrin, and eleutheroside E in botanical samples of Apocynum venetum L. and Eleutherococcus senticosus used in traditional Chinese medicine by HPLC combined with diode array detection (DAD)
  • Ionic liquid vacuum microwave-assisted extraction (ILVMAE) of rutin, hyperoside, and hesperidin from Sorbus tianschanica leaf samples for their subsequent quantification by HPLC
  • Simultaneous detection and quantification of vitexin, rutin, hyperoside, and quercetin in hawthorn flower and leaf extract samples using high-performance liquid chromatography combined with spectrophotometric detection (HPLC-UV)

Biochem/physiol Actions

Protects against peroxide-induced oxidative damage to cells by scavenging reactive oxygen species and enhancing activity of anti-oxidant enzymes, in particular, catalase and glutathione peroxidase.

Other Notes

A flavonol glycoside widely distribute in plants, including mimosa and St. John′s wort.
This compound is commonly found in plants of the genus: crataegus eucalyptus hypericum sambucus

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mao-Hua Ping
Cancer management and research, 12, 11773-11782 (2020-11-26)
Hyperin is an effective monomer extracted from Malvaceae plant Abelmoschus, which is a flavonol glycoside compound. Hyperin performs a variety of pharmacological activities, such as analgesia, antioxidation, anti-inflammation, avoiding microthrombosis, regulating immune function, inhibiting tumor cell growth. But the role
Sae-Kwang Ku et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 53, 197-204 (2012-12-12)
The potential anticoagulant activities of two single compounds, isorhamnetin-3-O-galactoside (IMG) and hyperoside, from Oenanthe javanica, were tested. The anticoagulant activities were investigated by measuring activated partial thromboplastin time (aPTT) and prothrombin time (PT), and the ability to inhibit production of
Rui-Li Liu et al.
Brain research, 1469, 164-173 (2012-07-10)
Hyperoside is a flavonoid compound and widely used in clinic to relieve pain and improve cardiovascular functions. However, the effects of hyperoside on ischemic neurons and the molecular mechanisms remain unclear. Here, we used an in vitro ischemic model of
Zi-lin Li et al.
Free radical biology & medicine, 57, 132-140 (2013-01-08)
One of the leading causes of death in the world is ischemia/reperfusion (I/R)-mediated acute myocardial infarction. There are a lot of Chinese traditional patent medicines, such as Xin'an capsules, Xin Xuening tablets, and so on, which have protective effects against
Zi-lin Li et al.
Journal of ethnopharmacology, 139(2), 388-394 (2011-11-29)
Hyperoside (Hyp) is a flavonoid compound isolated from Rhododendron ponticum L. leaves that elicits vascular protective effects in vitro. Treatment with Hyp has been found to attenuate endothelial cell damage induced by oxidative stress, but its mechanisms of action remain

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service