Merck
New and emerging cosmetic allergens.
Clinics in dermatology (2011-04-19)
Rosie F Davies, Graham A Johnston
ABSTRACT
Human skin is exposed to a large variety of cosmetic allergens. Most allergic contact dermatitis occurs after exposure to fragrance, preservatives, and hair dyes. Such reactions can often be occult. As a result, a high index of suspicion is needed in assessing the patient with facial or cosmetic dermatitis. This contribution looks at why such a large number of chemicals are in everyday usage, at how dermatologists monitor trends in allergy to cosmetics, and at a number of new and emerging allergens to consider in the assessment of suspected cosmetic allergy.
Sigma-Aldrich
Sodium metabisulfite, JIS first grade, ≥95.0%
Supelco
Sodium metabisulfite, analytical standard
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%
Sigma-Aldrich
Sodium metabisulfite, puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%
Sigma-Aldrich
Sodium metabisulfite, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
3-Iodo-2-propynyl N-butylcarbamate, 97%