Skip to Content
Merck
All Photos(3)

Key Documents

S9000

Sigma-Aldrich

Sodium metabisulfite

ReagentPlus®, ≥99%

Synonym(s):

Sodium disulfite, Sodium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O5
CAS Number:
Molecular Weight:
190.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

powder

pH

4.5 (20 °C, 50 g/L)

mp

>300 °C (lit.)

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

HRZFUMHJMZEROT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Sickle Erythrocyte Health Index: Discusses the use of sodium metabisulfite in stabilizing red blood cells for advanced computational analysis, crucial for the diagnosis and management of sickle cell disease (Ansong-Ansongton YON, Adamson TD, 2024).

  • Chemical and Thermal Treatment for Drying Cassava Tubers: Explores sodium metabisulfite′s role in enhancing the drying process of cassava, focusing on optimizing microstructure and dehydration kinetics, which is vital for food preservation and processing (Nainggolan EA, Banout J, Urbanova K, 2023).

  • Anionic Bio-flocculants from Sugarcane: Discusses the development of bio-flocculants using sodium metabisulfite for the purification of sucrose, demonstrating its potential in enhancing the sustainability of sugar processing industries (Leão S, Magalhães S, et al., 2023).

Other Notes

As a solid, this product is sodium metabisulfite, Na2S2O5. Dissolved in water, it yields NaHSO3.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Brelsford et al.
Current biology : CB, 30(2), 304-311 (2020-01-07)
Supergenes, clusters of tightly linked genes, play a key role in the evolution of complex adaptive variation [1, 2]. Although supergenes have been identified in many species, we lack an understanding of their origin, evolution, and persistence [3]. Here, we
Ivana Mara da Costa Machado Matos Carvalho et al.
Chemosphere, 82(7), 1056-1061 (2010-11-09)
Sodium metabisulfite is used in marine shrimp harvesting to prevent the occurrence of black spots. Shrimps are soaked in a sodium metabisulfite solution in ice, which is disposed of in sewages that run into marine canals, creating an environmental hazard.
Jeffrey N Hemenway et al.
Journal of pharmaceutical sciences, 101(9), 3305-3318 (2012-05-23)
A 2,4-dinitrophenylhydrazine (DNPH) precolumn derivatization high-performance liquid chromatography-ultraviolet detection (HPLC-UV) method was developed to quantify levels of formaldehyde and acetaldehyde in polyethylene glycol (PEG) solutions. Formic acid and acetic acid were quantified by HPLC-UV. Samples of neat and aqueous PEG
Gisela Palma-Orozco et al.
Phytochemistry, 72(1), 82-88 (2010-11-23)
While a long shelf life for fruit products is highly desired, enzymatic browning is the main cause of quality loss in fruits and is therefore a main problem for the food industry. In this study polyphenol oxidase (PPO), the main
Rosie F Davies et al.
Clinics in dermatology, 29(3), 311-315 (2011-04-19)
Human skin is exposed to a large variety of cosmetic allergens. Most allergic contact dermatitis occurs after exposure to fragrance, preservatives, and hair dyes. Such reactions can often be occult. As a result, a high index of suspicion is needed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service