Skip to Content
Merck
All Photos(2)

Key Documents

1.06357

SAFC

Sodium disulfite

(sodium metabisulfite), EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF, E 223

Pharma Manufacturing

Synonym(s):

Sodium disulfite, Sodium metabisulfite, Sodium pyrosulfite, Disulfite, Pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O5S2
CAS Number:
Molecular Weight:
190.11
MDL number:
UNSPSC Code:
12352302
E Number:
E223
EC Index Number:
231-673-0
Pricing and availability is not currently available.

Agency

BP
JP
NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

pH

3.5-5 (20 °C, 50 g/L in H2O)

mp

150 °C (decomposition)

density

2.36 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[S](=O)([O-])O[S](=O)[O-]

InChI

1S/2Na.H2O5S2/c;;1-6(2)5-7(3)4/h;;(H,1,2)(H,3,4)/q2*+1;/p-2

InChI key

LDTLADDKFLAYJA-UHFFFAOYSA-L

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium disulfite has reducing and anti-septic properties with broad range of applications in pharmaceutical, cosmetic and food industry. It is commonly used as preservative and antioxidant, especially for liquid and semi-solid application. In the food industry it is can help preserving food products and maintaining their fresh appearance.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service