Skip to Content
Merck
All Photos(5)

Key Documents

31448

Sigma-Aldrich

Sodium metabisulfite

puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%

Synonym(s):

Sodium disulfite, Sodium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O5
CAS Number:
Molecular Weight:
190.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
98-100.5%
form:
powder

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

Assay

98-100.5%

form

powder

quality

dry

impurities

≤0.01% thiosulfate (S2O3)

pH

3.5-5.0 (20 °C, 5%)

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2Na.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

HRZFUMHJMZEROT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium metabisulfite (MBS, Sodium disulfite) is an inorganic salt. It is the sodium salt of disulphurous (pyrosulfurous) acid. It is widely used in textile dyeing, photography and paper industry. It is also commonly added to various food products and wines as a preservative.

Application

Sodium metabisulfite may be used as an initiator during the cross-linking polymerization of polybutadiene in the cores of the vesicle membranes. It may be added as an antioxidant during the preparation of stock solutions of 6-hydroxydopamine in various studies.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanne Louise Neiiendam et al.
Journal of neurochemistry, 91(4), 920-935 (2004-11-05)
The Neural Cell Adhesion Molecule (NCAM) plays a crucial role in development of the central nervous system regulating cell migration, differentiation and synaptogenesis. NCAM mediates cell-cell adhesion through homophilic NCAM binding, subsequently resulting in activation of the fibroblast growth factor
W Wright et al.
The American review of respiratory disease, 141(6), 1400-1404 (1990-06-01)
Sodium metabisulfite (MBS) is used as a preservative in food and wine and frequently triggers attacks of asthma. To determine the characteristics of responses to inhaled MBS, 30 asthmatic subjects and 16 nonasthmatic subjects inhaled MBS, in concentrations of 6.2
Zhiliang Cheng et al.
Langmuir : the ACS journal of surfaces and colloids, 24(15), 8169-8173 (2008-06-24)
The ability of chelated Gd to serve as an effective magnetic resonance (MR) contrast agent largely depends on fast exchange rates between the Gd-bound water molecules and the surrounding bulk water. Because water diffuses slowly across lipid bilayers, liposomes with
Laurie H L Sellings et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 23(15), 6295-6303 (2003-07-18)
Convergent evidence suggests that amphetamine (AMPH) exerts its rewarding and locomotor stimulating effects via release of dopamine in the nucleus accumbens. However, there is no consensus as to the relative contributions of core and medial shell subregions to these effects.
Eagleson M.
Concise Encyclopedia Chemistry, 337-337 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service