Skip to Content
Merck
All Photos(1)

Key Documents

A-903

Supelco

Alprazolam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClN4
CAS Number:
Molecular Weight:
308.76
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cc1nnc2CN=C(c3ccccc3)c4cc(Cl)ccc4-n12

InChI

1S/C17H13ClN4/c1-11-20-21-16-10-19-17(12-5-3-2-4-6-12)14-9-13(18)7-8-15(14)22(11)16/h2-9H,10H2,1H3

InChI key

VREFGVBLTWBCJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alprazolam is a benzodiazepine drug used for the medical treatment of anxiety and panic disorders, including generalized or social anxiety disorder. This certified reference standard is suitable for use in a wide variety of LC/MS or GC/MS testing applications from forensic analysis and clinical toxicology to urine drug testing and pain prescription monitoring. Alprazolam is sold under the trade name Xanax®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Xanax is a registered trademark of Pharmacia & Upjohn Company

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dean T Acheson et al.
Human psychopharmacology, 27(4), 419-427 (2012-07-12)
Heightened anticipation of future events has been characterized as a feature of certain anxiety disorders. In functional magnetic resonance imaging studies, anticipation of fearful/threatening images has been shown to robustly activate the insular cortex and amygdala in healthy subjects, in
J M Jonas et al.
The Journal of clinical psychiatry, 54 Suppl, 25-45 (1993-10-01)
A review of the worldwide published literature was conducted to assess the efficacy and safety of alprazolam for the treatment of anxiety disorders, panic disorder, and depression in comparison with those of other active drugs (including other benzodiazepines and antidepressant
E M Sellers et al.
The Journal of clinical psychiatry, 54 Suppl, 64-75 (1993-10-01)
The incidence of nonmedical use of alprazolam is very low relative to its widespread legitimate medical use; in fact, given the millions of patients who have received this medication, the incidence is remarkably small. In particular, among patients with anxiety
J O Cole et al.
The Journal of clinical psychiatry, 54 Suppl, 49-61 (1993-10-01)
A review of the published case reports of adverse behavioral episodes or unexpected psychopathology in patients taking benzodiazepines was undertaken in an attempt to determine if these adverse or unexpected events are more likely to occur with alprazolam when compared
Meitong Zang et al.
Malaria journal, 13, 214-214 (2014-06-04)
Artequick is a relatively inexpensive artemisinin (Qing-hao-su; QHS)-based combination therapy (ACT) that contains QHS and piperaquine (PQ), which has not been widely used because of the decreased concentration level of QHS after repeated oral administrations for five to seven days

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service