Skip to Content
Merck
All Photos(1)

Documents

A0357000

Alprazolam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

8-Chloro-1-methyl-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClN4
CAS Number:
Molecular Weight:
308.76
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

alprazolam

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cc1nnc2CN=C(c3ccccc3)c4cc(Cl)ccc4-n12

InChI

1S/C17H13ClN4/c1-11-20-21-16-10-19-17(12-5-3-2-4-6-12)14-9-13(18)7-8-15(14)22(11)16/h2-9H,10H2,1H3

InChI key

VREFGVBLTWBCJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alprazolam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weam Elbezanti et al.
Viruses, 12(2) (2020-02-14)
Antiretroviral therapy (ART) lowers human immunodeficiency virus type 1 (HIV-1) viral load to undetectable levels, but does not eliminate the latent reservoir. One of the factors controlling the latent reservoir is transcriptional silencing of the integrated HIV-1 long terminal repeat
Dean T Acheson et al.
Human psychopharmacology, 27(4), 419-427 (2012-07-12)
Heightened anticipation of future events has been characterized as a feature of certain anxiety disorders. In functional magnetic resonance imaging studies, anticipation of fearful/threatening images has been shown to robustly activate the insular cortex and amygdala in healthy subjects, in
J O Cole et al.
The Journal of clinical psychiatry, 54 Suppl, 49-61 (1993-10-01)
A review of the published case reports of adverse behavioral episodes or unexpected psychopathology in patients taking benzodiazepines was undertaken in an attempt to determine if these adverse or unexpected events are more likely to occur with alprazolam when compared
E M Sellers et al.
The Journal of clinical psychiatry, 54 Suppl, 64-75 (1993-10-01)
The incidence of nonmedical use of alprazolam is very low relative to its widespread legitimate medical use; in fact, given the millions of patients who have received this medication, the incidence is remarkably small. In particular, among patients with anxiety
J M Jonas et al.
The Journal of clinical psychiatry, 54 Suppl, 25-45 (1993-10-01)
A review of the worldwide published literature was conducted to assess the efficacy and safety of alprazolam for the treatment of anxiety disorders, panic disorder, and depression in comparison with those of other active drugs (including other benzodiazepines and antidepressant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service