Skip to Content
Merck
All Photos(1)

Key Documents

N-906

Supelco

Nitrazepam solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O3
CAS Number:
Molecular Weight:
281.27
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1ccc2NC(=O)CN=C(c3ccccc3)c2c1

InChI

1S/C15H11N3O3/c19-14-9-16-15(10-4-2-1-3-5-10)12-8-11(18(20)21)6-7-13(12)17-14/h1-8H,9H2,(H,17,19)

InChI key

KJONHKAYOJNZEC-UHFFFAOYSA-N

General description

A certified solution standard suitable for use as starting material in calibrators and controls of nitrazepam for LC/MS or GC/MS testing methods. Nitrazepam is a benzodiazepine sold as a treatment for severe insomnia. This drug is marketed under the trade names Mogadon, Nitrados, and Somnite.

Application

  • Nitrazepam research solution in sedative pharmacological studies: Nitrazepam, a benzodiazepine, is extensively used in pharmacological research focusing on its sedative effects. Studies explore its impact on the central nervous system and its potential therapeutic applications for sleep disorders and anxiety (Bhoir et al., 1999).
  • Neuroscientific research applications of Nitrazepam: The solution is vital in neuroscientific studies, particularly those investigating the modulation of GABAergic systems by benzodiazepines. Nitrazepam is used to elucidate the mechanisms through which benzodiazepines affect neurotransmitter release and neuronal excitability (Honeychurch et al., 2006).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihalj Poša et al.
Colloids and surfaces. B, Biointerfaces, 86(2), 285-291 (2011-05-07)
The formation of mixed micelles built of 7,12-dioxolithocholic and the following hydrophobic bile acids was examined by conductometric method: cholic (C), deoxycholic (D), chenodeoxycholic (CD), 12-oxolithocholic (12-oxoL), 7-oxolithocholic (7-oxoL), ursodeoxycholic (UD) and hiodeoxycholic (HD). Interaction parameter (β) in the studied
Masatomo Suetsugi et al.
Progress in neuro-psychopharmacology & biological psychiatry, 31(4), 839-847 (2007-03-03)
The first-night effect is a well-known phenomenon that is considered to result from a subject's lack of adaptation to the unfamiliar environment of a sleep laboratory and to the technical equipment used for polysomnography. The effect has been explored as
Treatment of infantile spasms with sodium valproate followed by benzodiazepines.
Pongsakdi Visudhiphan
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 90(10), 2244-2244 (2007-11-29)
Mihalj Posa et al.
Colloids and surfaces. B, Biointerfaces, 74(1), 84-90 (2009-07-28)
Critical micellear concentrations (CMC) were determined for two novel promoters of membrane permeability-7-monoketocholic acid (7-MKC) and 12-monoketocholic acid (12-MKC), using two non-invasive ((1)H NMR relaxation experiment and conductometry) and two invasive (spectral shift and partition coefficient of the probe molecule)
Katsuhiko Mizuno et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(2), 345-351 (2008-11-14)
Cytochrome P450 3A4 is the predominant isoform in liver, and it metabolizes more than 50% of the clinical drugs commonly used. However, CYP3A4 is also responsible for metabolic activation of drugs, leading to liver injury. Benzodiazepines are widely used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service