Skip to Content
Merck
All Photos(1)

Documents

D-907

Supelco

Diazepam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClN2O
CAS Number:
Molecular Weight:
284.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CN1C(=O)CN=C(c2ccccc2)c3cc(Cl)ccc13

InChI

1S/C16H13ClN2O/c1-19-14-8-7-12(17)9-13(14)16(18-10-15(19)20)11-5-3-2-4-6-11/h2-9H,10H2,1H3

InChI key

AAOVKJBEBIDNHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Spiking Solution® is suitable for testing diazepam by HPLC, GC/MS or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and cannabis potency or impurity profiling testing applications.

Diazepam is an anxiolytic benzodiazepine drug, commonly used to treat anxiety disorders, muscle spasms, and acute seizures among others.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Electrochemical determination of diazepam in serum, urine, and tablet formulations by a modified electrode based on fullerene-functionalized carbon nanotubes and ionic liquid (C60-CNTs/IL)
  • Differential voltametric analysis of diazepam in its tablet formulations and human urine samples using a bismuth-modified pre-treated pencil graphite electrode (BiPPGE)
  • Simultaneous quantitative determination of diazepam and flunitrazepam in alcoholic beverage samples by Direct-electron ionization-liquid chromatography-tandem mass spectrometry (EI-LC-MS/MS) method
  • Dispersive nanomaterial-ultrasound assisted-microextraction (DNUM) of diazepam and chlordiazepoxide from urine and plasma samples for their quantification by high-performance liquid chromatography -UV (HPLC-UV) detection
  • Multi-residue analysis of diazepam and its metabolites in human oral fluid samples by their solid-phase extraction followed by LC-MS/MS method-based determination

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical determination of diazepam in real samples based on fullerene-functionalized carbon nanotubes/ionic liquid nanocomposite
Rahimi-Nasrabadi M, et al.
Sensors and Actuators B, Chemical, 240, 125-131 (2017)
Voltammetric determination of diazepam using a bismuth modified pencil graphite electrode
Dehghanzade M and Alipour E
Analytical Methods : Advancing Methods and Applications, 8, 1995-2004 (2016)
A Amiri Pebdani et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1008, 146-155 (2015-12-15)
Benzodiazepines (BDs) are used widely in clinical practice, due to their multiple pharmacological functions. In this study a dispersive nanomaterial-ultrasound assisted- microextraction (DNUM) method followed by high performance liquid chromatography (HPLC) was used for the preconcentration and determination of chlordiazepoxide
Berend Malchow et al.
Journal of neural transmission (Vienna, Austria : 1996), 122(7), 1019-1033 (2014-10-14)
Hippocampus volumes have been shown to be decreased in patients with major depression, but volume measurements are inconsistent in patients with bipolar disorder. Both disorders are associated with deficits in hippocampus-mediated cognitive functions. However, the underlying pathophysiology is widely unknown.
Haim Bassan et al.
Epilepsia, 54(6), 1092-1098 (2013-04-05)
Prolonged febrile seizures (PFS) lasting ≥15 min have been associated with increased risk for epilepsy in later life. Initial treatment, mostly prehospital, aims to prevent its evolution to febrile status epilepticus (FSE) and reduce adverse outcome. Paucity of information is

Articles

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service