Skip to Content
Merck
All Photos(1)

Key Documents

A-007

Supelco

(±)-Amphetamine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-Amphetamine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(N)Cc1ccccc1

InChI

1S/C9H13N/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3

InChI key

KWTSXDURSIMDCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs such as Adderall®, Vyvanse®, and ProCentra®. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This Certified Spiking Solution® is applicable for use in urine drug testing, sports testing, clinical toxicology, or forensic analysis applications by LC-MS/MS or GC/MS.

Legal Information

Adderall is a registered trademark of Richwood Pharmaceutical Company, Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
ProCentra is a registered trademark of Outlook Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyvanse is a registered trademark of Shire, LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily E Reichard et al.
Pharmaceutical research, 33(12), 2954-2966 (2016-11-04)
Methamphetamine (METH) abuse is a worldwide drug problem, yet no FDA-approved pharmacological treatments are available for METH abuse. Therefore, we produced an anti-METH single chain antibody fragment (scFv7F9Cys) as a pharmacological treatment for METH abuse. ScFv's have a short half-life
John F Bowyer et al.
BMC genomics, 14, 147-147 (2013-03-19)
The meninges (arachnoid and pial membranes) and associated vasculature (MAV) and choroid plexus are important in maintaining cerebrospinal fluid (CSF) generation and flow. MAV vasculature was previously observed to be adversely affected by environmentally-induced hyperthermia (EIH) and more so by
Prapimpun Wongchitrat et al.
Neuroscience letters, 542, 126-130 (2013-03-23)
Drug addicts have severe disruptions in many physiological and behavioral rhythms, such as the sleep/wake cycle. Interestingly, amphetamine, a psychostimulant, is able to alter many circadian patterns, which are independent of the master biological clock located in the suprachiasmatic nucleus.
Maria Xilouri et al.
Brain : a journal of neurology, 136(Pt 7), 2130-2146 (2013-06-13)
α-Synuclein levels are critical to Parkinson's disease pathogenesis. Wild-type α-synuclein is degraded partly by chaperone-mediated autophagy, and aberrant α-synuclein may act as an inhibitor of the pathway. To address whether the induction of chaperone-mediated autophagy may represent a potential therapy
An argument for permitting amphetamines and instant-release methylphenidate.
Jessica Flanigan
The American journal of bioethics : AJOB, 13(7), 49-51 (2013-06-19)

Protocols

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service