Skip to Content
Merck
All Photos(1)

Documents

C4895

Sigma-Aldrich

Cephalexin hydrate

first-generation cephalosporin antibiotic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O4S · xH2O
CAS Number:
Molecular Weight:
347.39 (anhydrous basis)
Beilstein:
965503
EC Number:
MDL number:
UNSPSC Code:
51282503
PubChem Substance ID:
NACRES:
NA.85

Quality Level

form

powder

pKa 

5.2
7.3

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

InChI

1S/C16H17N3O4S.H2O/c1-8-7-24-15-11(14(21)19(15)12(8)16(22)23)18-13(20)10(17)9-5-3-2-4-6-9;/h2-6,10-11,15H,7,17H2,1H3,(H,18,20)(H,22,23);1H2/t10-,11-,15?;/m1./s1

InChI key

AVGYWQBCYZHHPN-FNOHQHCYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Cephalexin is a cephalosporin antibiotic used to study the effect of expression, binding, and inhibition of PBP3 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis.

Biochem/physiol Actions

Cephalexin disrupts the synthesis of the peptidoglycan layer of bacterial cell walls which is responsible for cell wall structural integrity. Peptidoglycan synthesis is facilitated by transpeptidases known as penicillin-binding proteins (PBPs). PBPs bind to the D-Ala-D-Ala at the end of muropeptides (peptidoglycan precursors) to crosslink the peptidoglycan. Cephalexin antibiotics mimic the D-Ala-D-Ala site, thereby competitively inhibiting PBP crosslinking of peptidoglycan.

Other Notes

Storage of this product should be in airtight containers and protected from light.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Perniss et al.
Scientific reports, 8(1), 5681-5681 (2018-04-11)
Several species of the Gram-negative genus Bordetella are the cause of respiratory infections in mammals and birds, including whooping cough (pertussis) in humans. Very recently, a novel atypical species, Bordetella pseudohinzii, was isolated from laboratory mice. These mice presented no
William T Gray et al.
Cell, 177(6), 1632-1648 (2019-06-01)
The scaling of organelles with cell size is thought to be exclusive to eukaryotes. Here, we demonstrate that similar scaling relationships hold for the bacterial nucleoid. Despite the absence of a nuclear membrane, nucleoid size strongly correlates with cell size
Hiroshi Kodaira et al.
The Journal of pharmacology and experimental therapeutics, 339(3), 935-944 (2011-09-22)
This study investigated the impact of the active efflux mediated by P-glycoprotein (P-gp) and breast cancer resistance protein (Bcrp) at the blood-brain barrier (BBB) on the predictability of the unbound brain concentration (C(u,brain)) by the concentration in the cerebrospinal fluid
Steven D Coon et al.
American journal of physiology. Gastrointestinal and liver physiology, 305(10), G678-G684 (2013-09-28)
Glucose-dependent insulinotropic polypeptide (GIP) secreted from jejunal mucosal K cells augments insulin secretion and plays a critical role in the pathogenesis of obesity and Type 2 diabetes mellitus. In recent studies, we have shown GIP directly activates Na-glucose cotransporter-1 (SGLT1)
Tomoko Sugiura et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(6), 1181-1188 (2008-03-07)
Gastrointestinal (GI) absorption of certain therapeutic agents is thought to be mediated by solute carrier (SLC) transporters, although minimal in vivo evidence has been reported. Here, we show key roles of postsynaptic density 95/disk-large/ZO-1 (PDZ) domain-containing protein, PDZK1, as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service