Skip to Content
Merck
All Photos(1)

Documents

C4417

Supelco

Cefuroxime sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N4NaO8S
CAS Number:
Molecular Weight:
446.37
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical
veterinary

storage temp.

2-8°C

SMILES string

[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(N)=O)=C(N2C1=O)C([O-])=O)c3ccco3

InChI

1S/C16H16N4O8S.Na/c1-26-19-9(8-3-2-4-27-8)12(21)18-10-13(22)20-11(15(23)24)7(5-28-16(17)25)6-29-14(10)20;/h2-4,10,14H,5-6H2,1H3,(H2,17,25)(H,18,21)(H,23,24);/q;+1/p-1/b19-9-;/t10-,14-;/m1./s1

InChI key

URDOHUPGIOGTKV-JTBFTWTJSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cefuroxime is a semisynthetic cephalosporin, widely used as an antimicrobial agent, in veterinary medication for the treatment of cattle, goats, etc.

Application

Cefuroxime sodium salt has been used as a standard in the analysis of cefuroxime sodium salt residues in domestic sheep milk using penzym enzymatic test method. Cefuroxime may be used as an analytical reference standard for the quantificaton of the analyte in pharmaceutical preparations using high-performance liquid chromatography technique.
Cefuroxime sodium salt has been used as a standard in the analysis of cefuroxime sodium salt residues in domestic sheep milk using penzym enzymatic test method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High performance liquid chromatographic method for the quantitative analysis of cefuroxime in pharmaceutical preparations
Journal of the Chilean Chemical Society, 60(2), 2869-2871 (2015)
Detection limits of ?-lactam antibiotics in ewe milk by Penzym enzymatic test
Althaus.L.R, et al.
Journal of Food Protection, 64(11), 1844-1847 (2001)
Antibiotic and chemotherapy e-book, 86(2), 457-463 (2010)
Chang Liu et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(10), 831-836 (2014-09-18)
To compare the effects of topical nonsteroidal anti-inflammatory drugs on pupil dilation maintenance during phacoemulsification cataract surgery and quantify the relationships between pupil size change and nuclear hardness. This prospective randomized clinical observation study was single centered and double-masked. We
Zhenqiang Fang et al.
Journal of investigative surgery : the official journal of the Academy of Surgical Research, 27(6), 354-359 (2014-07-06)
Open nephroureterectomy (ONU) and bladder cuff resection (ONU-BCR) has been the gold standard of surgical treatment for upper urinary tract transitional cell carcinoma (UUT-TCC). The aim of this study is to introduce a modified total retroperitoneal laparoscopic nephroureterectomy (LNU) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service