Skip to Content
Merck
All Photos(1)

Documents

C6895

Sigma-Aldrich

Cefaclor

Synonym(s):

7-(D-2-Amino-2-phenylacetamido)-3-chloro-3-cephem-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14ClN3O4S
CAS Number:
Molecular Weight:
367.81
Beilstein:
8176092
EC Number:
MDL number:
UNSPSC Code:
51283701
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

Quality Level

solubility

soluble (slightly soluble in water, practically insoluble in methanol, ethanol)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

SMILES string

N[C@@H](C(=O)N[C@H]1[C@H]2SCC(Cl)=C(N2C1=O)C(O)=O)c3ccccc3

InChI

1S/C15H14ClN3O4S/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7/h1-5,9-10,14H,6,17H2,(H,18,20)(H,22,23)/t9-,10-,14-/m1/s1

InChI key

QYIYFLOTGYLRGG-GPCCPHFNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Cefaclor has been used in the heterologous expression of AaOr7 and AaOr8 in Xenopus laevis oocytes. It has also been used in cultures of isolated trigeminal ganglion (TG) and dorsal root ganglion (DRG) neurons.
Cefaclor is used to study urinary tract, intra-abdominal, and Haemophilus influenzae infections . It is used to study the mechanism of human renal organic anion and peptide transporters such as hOAT1, hPEPT1, and hPEPT2 and to study the effects of inhibition of penicillin-binding proteins on bacterial cell wall mucopeptide synthesis .

Biochem/physiol Actions

Cefaclor is a second generation cephalosporin with broad spectrum activity against Gram-negative and Gram-positive bacteria. It binds to penicillin-binding proteins and thereby inhibits the cell wall synthesis causing cell death.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Slightly soluble in water, practically insoluble in methanol, chloroform and benzene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiran Singh et al.
European journal of medicinal chemistry, 42(3), 394-402 (2007-01-17)
A few (1:1) and (1:2) metal complexes of cobalt(II), nickel(II), copper(II) and zinc(II) have been isolated with ligand derived from the condensation of 4-amino-3-mercapto-6-methyl-5-oxo-1,2,4-triazine with 2-acetylpyridine (L(1)) and characterized by elemental analysis, conductivity measurements, infrared, electronic, (1)H NMR spectral data
Lysophosphatidic acid-induced itch is mediated by signalling of LPA5 receptor, phospholipase D and TRPA1/TRPV1
Kittaka H, et al.
The Journal of Physiology, 595(8), 2681-2698 (2017)
Nikita B Ruparel et al.
Journal of endodontics, 38(10), 1372-1375 (2012-09-18)
Regenerative endodontic procedures are an alternative treatment for immature teeth with necrotic pulps. Typically, intracanal medicaments such as triple antibiotic paste (TAP) or double antibiotic paste (DAP) and calcium hydroxide (Ca[OH](2)) are used for disinfection. However, their effect on human
M Picard et al.
Antimicrobial agents and chemotherapy, 36(11), 2569-2572 (1992-11-01)
Cefaclor sustained its inhibitory activity against a beta-lactamase-producing strain of Haemophilus influenzae. Although a relatively high permeability coefficient was calculated for ampicillin compared with that calculated for cefaclor, the resulting periplasmic concentration of cefaclor was 5.7 times that of ampicillin.
Basma Dabbagh et al.
Pediatric dentistry, 34(5), 414-417 (2012-12-06)
The purpose of this case series was to report on the use of a technique of revascularization for necrotic immature permanent teeth, several problems encountered, and solutions to those problems. Eighteen pulp revascularizations were performed in 2009 using the original

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service