Skip to Content
Merck
All Photos(1)

Key Documents

K4013

Sigma-Aldrich

Kasugamycin hydrochloride from Streptomyces kasugaensis

Synonym(s):

3-O-[2-Amino-4-[(carboxyiminomethyl)amino]-2,3,4,6-tetradeoxy-D-arabino-hexopyranosyl]-D-chiro-inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25N3O9 · HCl
CAS Number:
Molecular Weight:
415.82
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

Streptomyces kasugaensis

Quality Level

form

powder

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)[C@H](O)[C@@H]2O)[C@@H](N)C[C@@H]1NC(C(O)=O)=N.Cl

InChI

1S/C14H25N3O9.ClH/c1-3-5(17-12(16)13(23)24)2-4(15)14(25-3)26-11-9(21)7(19)6(18)8(20)10(11)22;/h3-11,14,18-22H,2,15H2,1H3,(H2,16,17)(H,23,24);1H/t3-,4+,5+,6-,7+,8+,9-,10+,11+,14-;/m1./s1

InChI key

ZDRBJJNXJOSCLR-NZXABURVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Kasugamycin (Ksg) is an aminoglycoside antibiotic isolated from Streptomyces kasugaensis. It is used to study ribosome structure and interactions near the m26A m26A sequence[1]. It is used to study the mechanisms of kasugamycin resistance[2].

Biochem/physiol Actions

Kasugamycin binds within the mRNA channel of the 30S subunit between the universally conserved G926 and A794 nucleotides in 16S ribosomal RNA[2]. It inhibits protein synthesis and binding of aminoacyl-sRNA to the ribosomes in fungi and induces f-met-tRNA dissociation from the P-site of the 30S subunit.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koichi Inoue et al.
Journal of bacteriology, 189(23), 8510-8518 (2007-09-25)
A 16S rRNA methyltransferase, KsgA, identified originally in Escherichia coli is highly conserved in all living cells, from bacteria to humans. KsgA orthologs in eukaryotes possess functions in addition to their rRNA methyltransferase activity. E. coli Era is an essential
Ceshing Sheu et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 45(5), 478-484 (2010-06-01)
A high performance liquid chromatographic (HPLC) analysis method with an ultraviolet (UV) detector and an Aqua C18 (250 x 4.6 mm, Phenomenex) column were applied to analyze the antibiotic fungicide kasugamycin in water. An aromatic sulfonic acid spe column (Backerbond
Reiko Amikura et al.
Mechanisms of development, 122(10), 1087-1093 (2005-08-30)
In Drosophila, mitochondrially encoded ribosomal RNAs (mtrRNAs) form mitochondrial-type ribosomes on the polar granules, distinctive organelles of the germ plasm. Since a reduction in the amount of mtrRNA results in the failure of embryos to produce germline progenitors, or pole
Atsushi Yoshii et al.
Applied and environmental microbiology, 78(16), 5555-5564 (2012-06-05)
Kasugamycin (KSM), a unique aminoglycoside antibiotic, has been used in agriculture for many years to control not only rice blast caused by the fungus Magnaporthe grisea but also rice bacterial grain and seedling rot or rice bacterial brown stripe caused
Rachel Binet et al.
BMC microbiology, 9, 279-279 (2010-01-02)
rRNA adenine dimethyltransferases, represented by the Escherichia coli KsgA protein, are highly conserved phylogenetically and are generally not essential for growth. They are responsible for the post-transcriptional transfer of two methyl groups to two universally conserved adenosines located near the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service