Skip to Content
Merck
All Photos(1)

Key Documents

1099008

USP

Cephalexin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cephalexin monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O4S · H2O
CAS Number:
Molecular Weight:
365.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cephalexin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

S1[C@H]2N(C(=C(C1)C)C(=O)O)C(=O)[C@H]2NC(=O)[C@H](N)c3ccccc3.O

InChI

1S/C16H17N3O4S.H2O/c1-8-7-24-15-11(14(21)19(15)12(8)16(22)23)18-13(20)10(17)9-5-3-2-4-6-9;/h2-6,10-11,15H,7,17H2,1H3,(H,18,20)(H,22,23);1H2/t10-,11-,15-;/m1./s1

InChI key

AVGYWQBCYZHHPN-CYJZLJNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cephalexin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Cephalexin Hydrochloride
  • Cephalexin Tablets
  • Cephalexin Capsules
  • Cephalexin for Oral Suspension
  • Cephalexin Tablets for Oral Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Raff
Postgraduate medical journal, 59 Suppl 5, 32-39 (1983-01-01)
Cephalexin has had 12 years of extensive clinical usage in the management of respiratory tract and other infections. It is reliably absorbed from the gastrointestinal tract and reaches therapeutic levels in serum and tissues. Toxicity and adverse side effects are
R S Griffith
Postgraduate medical journal, 59 Suppl 5, 16-27 (1983-01-01)
Cephalexin has an antimicrobial spectrum that includes those pathogens most frequently encountered in clinical practice. It is useful in the treatment of infections of the upper and lower respiratory tract, skin and soft tissue, and the genitourinary tract. It can
R N Jones et al.
Postgraduate medical journal, 59 Suppl 5, 9-15 (1983-01-01)
The present report reviews the significant in vitro characteristics of cephalexin based on data published since 1967 (Wick, 1967). Whenever possible, current data on the action spectrum of cephalexin are compared with previous results, and the significance of in vitro
K M Murray et al.
The Annals of pharmacotherapy, 26(10), 1230-1233 (1992-10-01)
To report a case of cephalexin-induced Stevens-Johnson syndrome (SJS), a devastating adverse drug reaction that involves the entire skin surface and mucosal areas of the body. MEDLINE search (key terms cephalosporins, Stevens-Johnson syndrome, erythema multiforme, and systemic lupus erythematosus) and
G L McCloskey et al.
Cutis, 59(5), 251-254 (1997-05-01)
The ampicillin rash occurring in cases of infectious mononucleosis is well documented. Similar phenomena have also been observed with other antibiotics. The case of a patient with infectious mononucleosis treated with cephalexin who later showed a rash is presented and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service