Skip to Content
Merck
All Photos(4)

Key Documents

C4786

Supelco

Cefoxitin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N3NaO7S2
CAS Number:
Molecular Weight:
449.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

form

powder or granules

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

storage temp.

2-8°C

SMILES string

[Na+].CO[C@]2(NC(=O)Cc1cccs1)C3SCC(COC(N)=O)=C(N3C2=O)C([O-])=O

InChI

1S/C16H17N3O7S2.Na/c1-25-16(18-10(20)5-9-3-2-4-27-9)13(23)19-11(12(21)22)8(6-26-15(17)24)7-28-14(16)19;/h2-4,14H,5-7H2,1H3,(H2,17,24)(H,18,20)(H,21,22);/q;+1/p-1/t14-,16+;/m1./s1

InChI key

GNWUOVJNSFPWDD-XMZRARIVSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cefoxitin is a β-lactam that belongs to the group of cephamycin antibiotics and is considered a 2nd generation cephalosporin. It shows resistance to β-lactamases and is used to treat a broad spectrum of bacterial infections caused by gram-positive and gram-negative bacteria, such as bronchitis, pharyngitis, meningitis, and urinary tract infection among others. It is used for intravenous and intramuscular injections. It is found to be active against Bacteroides and is therapeutically used for the treatment of mixed aerobic-anaerobic infections.

Application

The analytical standard can be used to:
  • Determine cefoxitin sodium using four stability-indicating spectrophotometric methods in the presence of its alkaline degradation products in its pure form and pharmaceutical dosage forms
  • Separate and measure cefoxitin sodium in commercial drug products by high-performance liquid chromatography (HPLC)

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrofluorimetric analysis of cefoxitin in pharmaceutical dosage
Talanta, 41(4), 557-563 (1994)
Methicillin-resistant Staphylococcus aureus screening by online immunometric monitoring of bacterial growth under selective pressure.
Stenholm T, et al.
Antimicrobial Agents and Chemotherapy, 53(12), 5088-5094 (2009)
Antibiotic and chemotherapy e-book, 14(3), 257-266 (2010)
Analysis of binary mixtures of cephalothin and cefoxitin by using first-derivative spectrophotometry
Murillo.A.J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 14(3), 257-266 (1996)
Matthew P Kronman et al.
Pediatrics, 130(4), e794-e803 (2012-09-26)
To determine whether childhood antianaerobic antibiotic exposure is associated with the development of inflammatory bowel disease (IBD). This retrospective cohort study employed data from 464 UK ambulatory practices participating in The Health Improvement Network. All children with ≥ 2 years

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service