Skip to Content
Merck
All Photos(1)

Documents

537551

Sigma-Aldrich

Ethylene glycol butyl ether

≥99%

Synonym(s):

2-Butoxyethanol, Butyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732511
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)

Assay

≥99%

form

liquid

autoignition temp.

473 °F

expl. lim.

10.6 %

impurities

≤0.10% (water)

refractive index

n20/D 1.419 (lit.)

bp

169-172.5 °C (lit.)

mp

−75 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCO

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3

InChI key

POAOYUHQDCAZBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol butyl ether (EGBE) is an ethylene oxide-based glycol ether solvent commonly used in the coatings industry. Its relative evaporation rate with respect to n-butyl acetate has been reported to be 0.07. Its toxicity has been assessed. The effect of EGBE as a thickening solvent during acrylic latex synthesis has been investigated.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - Pensky-Martens closed cup

Flash Point(C)

67 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel alkali and cosolvent thickening mechanism for latex.
He L, et al.
New. J. Chem., 39(11), 8984-8992 (2015)
R L Smith
Environmental health perspectives, 57, 1-4 (1984-08-01)
Ethylene oxide-based glycol ether and glycol ether ester solvents have been used in the coatings industry for the past fifty years. Because of their excellent performance properties (evaporation rate, blush resistance, flow-out and leveling properties, solubility for coating resins, solvent
Reproductive toxicology. Ethylene glycol monobutyl ether.
Environmental health perspectives, 105 Suppl 1, 217-218 (1997-02-01)
Ryszard Tokarczyk et al.
Journal of chromatography. A, 1217(44), 6964-6970 (2010-09-22)
A gas chromatography-mass spectrometry isotope dilution (GC-MS ID) method was developed and tested for the determination of 14 common glycol ethers in consumer products. Stable isotope labelled standards, 2-methoxyethanol-D(7) and 2-butoxyethanol-(13)C(2) (CDN isotopes) were employed to enhance the accuracy and
Ilona Sperlingová et al.
Analytical and bioanalytical chemistry, 397(2), 433-438 (2009-10-02)
Ethylene glycol monobutyl ether (EGBE), an industrial solvent, is absorbed by the body not only by inhalation but also by dermal absorption (liquid or vapour). EGBE is metabolized to butoxyacetic acid (BAA). Pooled freeze-dried urine candidate reference material (RM) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service