Skip to Content
Merck
All Photos(1)

Key Documents

307289

Sigma-Aldrich

2-Butoxyethyl acetate

99%

Synonym(s):

1-Acetoxy-2-butoxyethane, BCA, Ethylene glycol monobutyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
160.21
Beilstein:
1756960
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.5 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

644 °F

expl. lim.

0.88 %, 33 °F
8.54 %, 135 °F

refractive index

n20/D 1.413 (lit.)

bp

192 °C (lit.)

density

0.942 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCOC(C)=O

InChI

1S/C8H16O3/c1-3-4-5-10-6-7-11-8(2)9/h3-7H2,1-2H3

InChI key

NQBXSWAWVZHKBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Butoxyethyl acetate is one of the components of Jiashi melon juice and was identified by gas chromatography-mass spectrometry-olfactometry (GC-MS-O) analysis. It stimulated the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes.

Application

  • Three-Dimensional Printing of Scaffolds: 2-Butoxyethyl acetate was utilized in the synthesis of poly(glycerol sebacate) acrylate for 3D printing applications, highlighting its role as a solvent in developing biocompatible materials for medical implants and tissue engineering (Wu et al., 2020).
  • Migration Analysis in Food Contact Materials: Demonstrates the application of 2-Butoxyethyl acetate in the extraction and analysis of migrating compounds from plastic baby bottles, using GC-MS to ensure consumer safety and compliance with food safety regulations (Onghena et al., 2014).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J N Lawrence et al.
Toxicology in vitro : an international journal published in association with BIBRA, 11(5), 627-631 (1997-10-01)
Damage to the skin induced by chemical irritants is associated with the release of arachidonic acid metabolites, such as prostaglandin E(2) (PGE(2)) which plays an important role in epidermal inflammation. This study investigated cytotoxicity and release of PGE(2) in human
Xueli Pang et al.
Journal of agricultural and food chemistry, 60(17), 4179-4185 (2012-04-14)
To identify aromatic compounds in Jiashi melon juice, gas chromatography-mass spectrometry-olfactometry (GC-MS-O) analysis was used. Odor activity values (OAVs) were also calculated on the basis of the qualitative and quantitative analysis of volatile compounds. Results showed that 42 volatiles were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service