Skip to Content
Merck
All Photos(3)

Documents

310328

Sigma-Aldrich

Propylene carbonate

anhydrous, 99.7%

Synonym(s):

1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
Beilstein:
107913
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor pressure

0.13 mmHg ( 20 °C)
0.98 mmHg ( 50 °C)

Assay

99.7%

form

liquid

autoignition temp.

851 °F

expl. lim.

14.3 %

impurities

<0.002% water
<0.005% water (100 mL pkg)

refractive index

n20/D 1.421 (lit.)

pH

7 (20 °C, 200 g/L)

bp

240 °C (lit.)

mp

−55 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

SMILES string

CC1COC(=O)O1

InChI

1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3

InChI key

RUOJZAUFBMNUDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propylene carbonate can be synthesized from propylene oxide and CO2. Optically active form of propylene carbonate can be prepared from the reaction between CO2 and racemic epoxides. Decomposition of propylene carbonate on the graphite electrode in lithium batteries results in the formation of a lithium intercalated compound.
Propylene carbonate is a cyclic carbonate that is commonly used as a solvent and as a reactive intermediate in organic synthesis. It is being considered as a potential electrochemical solvent due to its low vapor pressure, high dielectric constant and high chemical stability.

Application

Propylene carbonate may be used as a solvent for the asymmetric hydrogenation of nonfunctionalized olefins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael L Aubrey et al.
Nature materials, 17(7), 625-632 (2018-06-06)
Conductive metal-organic frameworks are an emerging class of three-dimensional architectures with degrees of modularity, synthetic flexibility and structural predictability that are unprecedented in other porous materials. However, engendering long-range charge delocalization and establishing synthetic strategies that are broadly applicable to
The cathodic decomposition of propylene carbonate in lithium batteries.
Arakawa M and Yamaki JI.
Journal of Electroanalytical Chemistry, 219(1-2), 273-280 (1987)
Vincenza Modafferi et al.
Nanomaterials (Basel, Switzerland), 10(8) (2020-08-19)
The effect of the type of dopant (titanium and manganese) and of the reduced graphene oxide content (rGO, 30 or 50 wt %) of the α-Fe2O3@rGO nanocomposites on their microstructural properties and electrochemical performance was investigated. Nanostructured composites were synthesized
Propylene carbonate: a versatile solvent for electrochemistry and EPR.
Nelson RF & Adams RN.
J. Electroanal. Chem. Interfac. Electrochem., 13(1), 184-187 (1967)
Xiao-Bing Lu et al.
Journal of the American Chemical Society, 126(12), 3732-3733 (2004-03-25)
This communication describes a convenient route to optically active propylene carbonate by a catalytic kinetic resolution process resulting from the coupling reaction of CO2 and racemic epoxides using simple chiral SalenCo(III)/quaternary ammonium halide catalyst systems.

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service