Skip to Content
Merck
All Photos(2)

Key Documents

484326

Sigma-Aldrich

Propylene glycol propyl ether

≥98.5%

Synonym(s):

DOWANOL® PnP, 1-Propoxy-2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OCH2CH(OH)CH3
CAS Number:
Molecular Weight:
118.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98.5%

refractive index

n20/D 1.411 (lit.)

bp

140-160 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

SMILES string

CCCOCC(C)O

InChI

1S/C6H14O2/c1-3-4-8-5-6(2)7/h6-7H,3-5H2,1-2H3

InChI key

FENFUOGYJVOCRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Dow Chemical
DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

115.5 °F - closed cup

Flash Point(C)

46.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ballantyne et al.
Veterinary and human toxicology, 30(2), 126-129 (1988-04-01)
1-Propoxy-2-propanol, a widely used industrial chemical, was found to have acute peroral LD50 values in the rat of 4.92 ml/kg (males) and 2.83 ml/kg (females), with the signs of systemic toxicity being principally related to narcosis. Acute percutaneous LD50 values
Sambhav R Kumbhani et al.
Rapid communications in mass spectrometry : RCM, 31(19), 1659-1668 (2017-08-07)
Ambient ionization mass spectrometry methods are convenient, sensitive and require little sample preparation. However, they are susceptible to species present in air surrounding the mass spectrometer. This study identifies some challenges associated with the potential impacts of indoor air contaminants
Yanqin Gao et al.
The Journal of chemical physics, 142(21), 214505-214505 (2015-06-08)
The dielectric relaxations in six primary and secondary alkoxy alcohols with varying molecular size and different separation between -O- and hydroxyl group are studied at temperatures around glass transition. The analyses of the apparent full width at half maximum of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service