Skip to Content
Merck
All Photos(1)

Key Documents

03351

Supelco

Diethylene glycol monobutyl ether

≥99.0%

Synonym(s):

2-(2-Butoxyethoxy)ethanol, BDG, Butyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
162.23
Beilstein:
1739225
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21
bp:
224-228 °C (lit.)
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (GC)
≥99.0%

form

liquid

impurities

≤0.1% water

refractive index

n20/D 1.432

bp

224-228 °C (lit.)

density

0.953 g/mL at 20 °C (lit.)

SMILES string

CCCCOCCOCCO

InChI

1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3

InChI key

OAYXUHPQHDHDDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Optimal Design of Alkaline-Surfactant-Polymer Flooding under Low Salinity Environment: Research highlights the use of diethylene glycol monobutyl ether in enhancing the efficiency of enhanced oil recovery methods, particularly in low salinity environments. This application points to its potential in reducing the environmental footprint of oil extraction processes (Novriansyah A, et al., 2020).

  • Bioaugmentation for treatment of full-scale diethylene glycol monobutyl ether (DGBE) wastewater by Serratia sp. BDG-2: Demonstrates the solvents biodegradability and the potential for its removal from wastewater using specific bacterial strains, highlighting important environmental safety considerations (Chen M, et al., 2016).

  • Diethylene glycol mono butyl ether concentrations in room air from application of cleaner formulations to hard surfaces: Examines the air quality implications of using diethylene glycol monobutyl ether in cleaning products, stressing the importance of monitoring and managing indoor air pollutants in industrial and domestic environments (Gibson WB, et al., 1991).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Göen et al.
Toxicology letters, 134(1-3), 295-300 (2002-08-23)
The aim of the study was to investigate the suitability of butoxyethoxyacetic acid (BEAA) as a biomarker of exposure to water-based cleaning agents containing diethylene glycol mono butyl ether (DEGBE). The study was performed in two printing plants where water-based
Yeomin Yoon et al.
Journal of membrane science, 261(1-2), 76-86 (2005-09-02)
Rejection characteristics of organic and inorganic compounds were examined for six reverse osmosis (RO) membranes and two nanofiltration (NF) membranes that are commercially available. A batch stirred-cell was employed to determine the membrane flux and the solute rejection for solutions
M Ema et al.
Drug and chemical toxicology, 11(2), 97-111 (1988-06-01)
The teratogenicity of diethylene glycol mono-n-butyl ether (DEGMBE) was studied in Wistar rats. The pregnant rats were fed a diet containing DEGMBE from day 0 through day 20 of pregnancy. The dietary concentrations of DEGMBE were 0, 0.04, 0.2 and
Christopher R Gibson et al.
Journal of the American Society for Mass Spectrometry, 14(11), 1247-1249 (2003-11-05)
Tandem liquid chromatography-mass spectrometry coupled to online radioactive material detection (LC/RAM/MS/MS) is a technique that is used routinely for in vivo and in vitro drug metabolism studies and allows for a simultaneous correlation between radiochemical peaks and mass spectral data.
W B Gibson et al.
Journal of exposure analysis and environmental epidemiology, 1(3), 369-383 (1991-07-01)
Diethylene glycol monobutyl ether (DGBE) is a solvent used in some liquid hard surface cleaners. We evaluated the inhalation component of consumer exposure in the home to DGBE from the use of cleaning products containing up to 9% DGBE. Several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service