256366
Ethylene glycol butyl ether
spectrophotometric grade, ≥99.0%
Synonym(s):
2-Butoxyethanol, Butyl glycol
About This Item
grade
spectrophotometric grade
vapor density
4.1 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)
Assay
≥99.0%
form
liquid
autoignition temp.
473 °F
expl. lim.
10.6 %
technique(s)
UV/Vis spectroscopy: suitable
impurities
≤0.1% water
evapn. residue
<0.001%
refractive index
n20/D 1.419 (lit.)
bp
169-172.5 °C (lit.)
mp
−75 °C (lit.)
density
0.902 g/mL at 25 °C (lit.)
λ
H2O reference
UV absorption
λ: 230 nm Amax: 1.0
λ: 250 nm Amax: 0.10
λ: 275 nm Amax: 0.05
λ: 300-400 nm Amax: 0.01
SMILES string
CCCCOCCO
InChI
1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3
InChI key
POAOYUHQDCAZBD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
152.6 °F - Pensky-Martens closed cup
Flash Point(C)
67 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service