Skip to Content
Merck
All Photos(1)

Key Documents

T9284

Sigma-Aldrich

Triton X-100

BioXtra

Synonym(s):

t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

description

non-ionic

product line

BioXtra

form

liquid

mol wt

micellar avg mol wt 80,000
average mol wt 625

aggregation number

100-155

technique(s)

permeability assay: suitable

impurities

≤0.05% Phosphorus (P)

ign. residue

≤0.4%

CMC

0.2-0.9 mM (20-25°C)

transition temp

cloud point 65 °C
pour point ~7 °C

solubility

water: soluble

density

1.065 g/cm3 at 20 °C (68 °F)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.0005%

HLB

13.5

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-100 is a common non-ionic surfactant and emulsifier which is often used in biochemical applications to solubilize proteins. It is considered a comparatively mild detergent, non-denaturing, and is reported in numerous references as a routinely added reagent. It is utilized for lysing cells to extract protein and cellular organelles. It can also permeabilize the living cell membrane for transfection.

Application

Triton X-100 has been used:
  • Along with Span® 80 and Tween® 80 in mineral oil for the preparation of oil-phase of microemulsions for PCR technique
  • As a component of NX buffer which is used to pool and break down the microemulsions after PCR cycling
  • In the blocking solution during immunocytochemistry procedure
  • During deglycosylation procedure
  • For membrane solubilization
  • For cell permeabilization
  • In decellularization of porcine heart valve tissue

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Features and Benefits

  • BioXtra Grade product for your Cell Biology and Biochemical research
  • Non-ionic nondenaturing detergent
  • Superior wetting agent and emulsifier
  • Highly versatile surfactant

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Span is a registered trademark of Croda International PLC
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proinflammatory chemokine induction in keratocytes and inflammatory cell infiltration into the cornea.
Hong JW et al.
Investigative Ophthalmology & Visual Science, 42, 2795-2795 (2001)
Angiotensin II AT1 receptor blockade decreases vasopressin-induced water reabsorption and AQP2 levels in NaCl-restricted rats.
Kwon TH et al.
American Journal of Physiology: Renal Physiology, 288, F673-F673 (2005)
Transforming single DNA molecules into fluorescent magnetic particles for detection and enumeration of genetic variations.
Dressman D, et al.
Proceedings of the National Academy of Sciences of the USA, 100(15), 8817-8822 (2003)
Structural, metabolic and developmental evaluation of ovulated rabbit oocytes before and after cryopreservation by vitrification and slow freezing.
Theriogenology, 74(5), 847-855 (2010)
Detection of fetal DNA in maternal plasma by microarray coupled with emulsions PCR.
Ge Q, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 369(1), 82-88 (2006)

Protocols

Cell staining can be divided into four steps: cell preparation, fixation, application of antibody, and evaluation.

Cell staining can be divided into four steps: cell preparation, fixation, application of antibody, and evaluation.

Cell staining can be divided into four steps: cell preparation, fixation, application of antibody, and evaluation.

Cell staining can be divided into four steps: cell preparation, fixation, application of antibody, and evaluation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service