Skip to Content
Merck
All Photos(1)

Key Documents

D5796

Sigma-Aldrich

DMEM - high glucose

Dulbecco’s Modified Eagle’s Medium - high glucose

With sodium Bicarbonate and ʟ-glutamine, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: yes
L-glutamine: yes
glucose: high
phenol red: yes
sodium pyruvate: no
HEPES: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This DMEM-Hi glucose medium is a 1x complete medium with no added factors (common modifications) such as L-alanyl-L-glutamine, HEPES, or sodium pyruvate. It differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Refael Minnes et al.
Scientific reports, 7(1), 4381-4381 (2017-07-01)
The vast majority of cancer related deaths are caused by metastatic tumors. Therefore, identifying the metastatic potential of cancer cells is of great importance both for prognosis and for determining the correct treatment. Infrared (IR) spectroscopy of biological cells is
Christian Kirschneck et al.
Scientific reports, 7(1), 14751-14751 (2017-11-09)
Meaningful, reliable and valid mRNA expression analyses by real-time quantitative PCR (RT-qPCR) can only be achieved, if suitable reference genes are chosen for normalization and if appropriate RT-qPCR quality standards are met. Human periodontal ligament (hPDL) fibroblasts play a major
Saabah B Mahbub et al.
Scientific reports, 7(1), 15792-15792 (2017-11-19)
Bioimaging using endogenous cell fluorescence, without any external biomarkers makes it possible to explore cells and tissues in their original native state, also in vivo. In order to be informative, this label-free method requires careful multispectral or hyperspectral recording of
Tomoyuki Muroi et al.
Reproduction (Cambridge, England), 154(6), 755-764 (2017-09-16)
G-protein-coupled receptors (GPCRs) participate in diverse physiological functions and are promising targets for drug discovery. However, there are still over 140 orphan GPCRs whose functions remain to be elucidated.
Kai-Liang Zhang et al.
OncoTargets and therapy, 12, 6515-6523 (2019-10-17)
The efficacy of traditional therapies for oral carcinoma (OC) is limited. Oncolytic adenovirus, a novel strategy of cancer therapy, shows potential use in OC treatment. However, its clinical application is limited by pre-existing neutralizing antibodies. Thus, this study aimed to

Protocols

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

Detailed procedure for how to perform a lentiviral transduction of MISSION shRNA lentiviral particles to achieve a stable long term silencing and phenotypic change.

See All

Related Content

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Questions

1–3 of 3 Questions  
  1. What are the product numbers for DMEM and EMEM used to grow the 293 and 293T cell lines?

    1 answer
    1. To grow the 293 and 293T cells, the following product recommendations are provided:

      For the 293 cell line (product number 85120602), ECACC recommends using EMEM media (EBSS) with 2mM Glutamine, 1% Non Essential Amino Acids (NEAA), and 10% FCS. ECACC uses the product number M4655 for this purpose.

      For the 293T cell line (product number 12022001), ECACC recommends using DMEM media with 2mM Glutamine and 10% Fetal Bovine Serum (FBS). ECACC uses the product number D5796 media for sub-culturing this cell line.

      Helpful?

  2. What is the conc. of L glutamine

    1 answer
    1. The concentration of L-Glutamine in this product is 0.584 g/L. Please see the link below to review the DMEM Formulation Tables: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/340/109/d5796for.pdf

      Helpful?

  3. pH는 몇으로 조정되어 있나요?

    1 answer
    1. The pH specification of this product is 7.0-7.6. The exact value will be reported in the lot specific Certificate of Analysis. Please see the link below to review a sample Certificate of Analysis: https://www.sigmaaldrich.com/coa/SIGMA/D5796/RNBL9771.

      Helpful?

Reviews

Active Filters

  1. Columbia, MO
    • Reviews 2
    • Vote 1
    5 out of 5 stars.

    DMEM

    I always feel confident when using Sigma cell culture media.

    Helpful?

  2. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service