Skip to Content
Merck
All Photos(3)

Documents

S9378

Sigma-Aldrich

Sucrose

≥99.5% (GC)

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

sugar cane

Assay

≥99.5% (GC)

form

crystals

technique(s)

gas chromatography (GC): suitable

impurities

≤5 ppm heavy metals

color

white

useful pH range

5.5-7.5 (25 °C, 342 g/L)

mp

185-187 °C (lit.)

solubility

H2O: 0.5 g/mL, clear, colorless

absorption

≤0.2 at 260 in water (50 G plus 50 mL)
≤0.2 at 280 in water (50 G plus 50 mL)

application(s)

agriculture

foreign activity

RNase, none detected

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sucrose has been used:
  • in plant cultivation and regeneration
  • cryopreservation
  • clear unobstructed brain/body imaging cocktails and computational analysis (CUBIC)

Biochem/physiol Actions

Sucrose, a non-reducing disaccharide is abundantly present in photosynthetic organisms including plants. It is the main constituent of molasses. Sucrose is used in food industry as liquid sugar. It is also a component of foodstuffs and beverages. In higher plants, sucrose participates in growth and development. It is an important human macronutrient. Sucrose is used in foods as a sweetening agent.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conductometric biosensors
Biological Identification, 153-193 (2014)
High-resolution 3D imaging of whole organ after clearing: taking a new look at the zebrafish testis
Fretaud M, et al.
Scientific reports, 7(8), 43012-43012 (2017)
Disaccharides, oligosaccharides and polysaccharides
Carbohydrates: The Essential Molecules of Life, 321-341 (2009)
Transformation of nuclear and plastomic plant genomes by biolistic particle bombardment
Maenpaa P, et al.
Molecular Biotechnology, 13(1), 67-72 (1999)
Ovary and Ovulation: Fresh and cryopreserved ovarian tissue samples from donors with lymphoma transmit the cancer to graft recipients
Shaw J M, et al.
Human Reproduction, 11(8), 1668-1673 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service