Skip to Content
Merck
All Photos(1)

Documents

R8758

Sigma-Aldrich

RPMI-1640 Medium

With ʟ-glutamine, sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: yes
phenol red: yes
sodium pyruvate: no
HEPES: no
L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.
RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Application

RPMI-1640 Medium has been used for maintaining cell line medium and serves as a medium for different cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E L Davies et al.
Clinical and experimental immunology, 145(1), 183-189 (2006-06-24)
An increasing number of cell types, including peripheral blood mononuclear cells (PBMCs), have been demonstrated to release heat shock proteins (Hsps). In this paper we investigate further the hypothesis that Hsps are danger signals. PBMCs and Jurkat cells released Hsp70
Katarzyna A Podyma-Inoue et al.
Cancer genomics & proteomics, 13(6), 443-452 (2016-11-04)
Heparan sulfate proteoglycans (HSPGs)-dependent endocytic events have been involved in glioma progression. Thus, comprehensive understanding of the intracellular trafficking complexes formed in presence of HSPGs would be important for development of glioma treatments. Subcellular fractionation was used to separate vesicles
Anders Abildgaard et al.
Psychoneuroendocrinology, 79, 40-48 (2017-03-05)
The gut microbiota has recently emerged as an important regulator of brain physiology and behaviour in animals, and ingestion of certain bacteria (probiotics) therefore appear to be a potential treatment for major depressive disorder (MDD). However, some conceptual and mechanistical
Michelle J Nyhan et al.
BMC cancer, 16, 101-101 (2016-02-18)
Successful treatment of oesophageal cancer is hampered by recurrent drug resistant disease. We have previously demonstrated the importance of apoptosis and autophagy for the recovery of oesophageal cancer cells following drug treatment. When apoptosis (with autophagy) is induced, these cells
Samira Khiar et al.
Scientific reports, 7(1), 2561-2561 (2017-06-02)
The type I interferon response plays a pivotal role in host defense against infectious agents and tumors, and promising therapeutic approaches rely on small molecules designed to boost this system. To identify such compounds, we developed a high-throughput screening assay

Protocols

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

Preparation for biodegradable nanoparticles and their use in transfection protocols .

See All

Related Content

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service